This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00384078_m1
See other MYDGF GE Assays ›
Gene Symbol:
MYDGF
Gene Name:
myeloid derived growth factor
Gene Aliases:
C19orf10, EUROIMAGE1875335, IL25, IL27, IL27w, R33729_1, SF20
Chromosome Location:
Chr.19: 4657545 - 4670403 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00384078_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MYDGF
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019107.3 NP_061980.1 3 - 4 354 80
GenBank mRNA AK098829.1 - 3 - 4 293 80
AL365373.1 - 3 - 4 276 80
AL365374.1 - 3 - 4 276 80
BC003639.2 - 3 - 4 272 80
BC010129.2 - 3 - 4 321 3953027 80
BC014655.1 - 3 - 4 292 4562455 80
BG484758.1 - 3 - 4 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1900009357.hg.1
This assay targets the same location as HTA transcript cluster(s) TC19001067.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MYDGF

Panther Classification:

Molecular Function -

growth factor

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
positive regulation of protein phosphorylation
positive regulation of endothelial cell proliferation
apoptotic process
positive regulation of phosphatidylinositol 3-kinase signaling
IRE1-mediated unfolded protein response
negative regulation of apoptotic process
positive regulation of MAPK cascade
positive regulation of angiogenesis
positive regulation of transcription from RNA polymerase II promoter
positive regulation of protein kinase B signaling
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR19014599.hg.1 ENST00000262947.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19014599.hg.1 NM_019107.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19014599.hg.1 TC19001067.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000596031.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000596031.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000596031.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000596031.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000596031.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000596031.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000599630.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000599630.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000599630.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000599630.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000599630.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000599630.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000599630.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000599761.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000599761.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000599761.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000599761.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 NM_019107.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 NM_019107.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 NM_019107.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 NM_019107.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 NM_019107.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 NM_019107.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 NM_019107.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 NM_019107.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 NM_019107.3 Refseq
Affymetrix Human Clariom D Assay PSR1900174774.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Clariom D Assay PSR1900174774.hg.1 ENST00000596031.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900174774.hg.1 ENST00000599630.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900174774.hg.1 ENST00000599761.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900174774.hg.1 NM_019107.3 Refseq
Affymetrix Human Clariom D Assay PSR1900174774.hg.1 TC1900009357.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900174774.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Clariom D Assay PSR1900174774.hg.1 ENST00000596031.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900174774.hg.1 ENST00000599630.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900174774.hg.1 ENST00000599761.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900174774.hg.1 NM_019107.3 Refseq
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000596031.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000596031.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000596031.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000596031.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000596031.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000596031.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000599630.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000599630.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000599630.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000599630.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000599630.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000599630.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000599630.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000599761.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000599761.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000599761.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000599761.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 NM_019107.3 Refseq
Affymetrix Human Clariom D Assay TC1900009357.hg.1 NM_019107.3 Refseq
Affymetrix Human Clariom D Assay TC1900009357.hg.1 NM_019107.3 Refseq
Affymetrix Human Clariom D Assay TC1900009357.hg.1 NM_019107.3 Refseq
Affymetrix Human Clariom D Assay TC1900009357.hg.1 NM_019107.3 Refseq
Affymetrix Human Clariom D Assay TC1900009357.hg.1 NM_019107.3 Refseq
Affymetrix Human Clariom D Assay TC1900009357.hg.1 NM_019107.3 Refseq
Affymetrix Human Clariom D Assay TC1900009357.hg.1 NM_019107.3 Refseq
Affymetrix Human Clariom D Assay TC1900009357.hg.1 NM_019107.3 Refseq
Affymetrix Human Clariom D Assay PSR1900174773.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Clariom D Assay PSR1900174773.hg.1 ENST00000596031.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900174773.hg.1 ENST00000599630.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900174773.hg.1 ENST00000599761.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900174773.hg.1 NM_019107.3 Refseq
Affymetrix Human Clariom D Assay PSR1900174773.hg.1 TC1900009357.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900174773.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Clariom D Assay PSR1900174773.hg.1 ENST00000596031.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900174773.hg.1 ENST00000599630.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900174773.hg.1 ENST00000599761.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900174773.hg.1 NM_019107.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline