This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00385005_m1
See other EPB41L1 GE Assays ›
Gene Symbol:
EPB41L1
Gene Name:
erythrocyte membrane protein band 4.1 like 1
Gene Aliases:
4.1N, MRD11
Chromosome Location:
Chr.20: 36064891 - 36232799 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00385005_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EPB41L1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001258331.1 NP_001245260.1 15 - 16 2133 57
NM_177996.2 NP_818932.1 15 - 16 2128 57
XM_011528683.1 XP_011526985.1 15 - 16 2196 57
XM_017027714.1 XP_016883203.1 15 - 16 2187 57
XM_017027720.1 XP_016883209.1 16 - 17 2010 57
GenBank mRNA AK299817.1 - 16 - 17 2300 57
BC013885.2 - 15 - 16 2133 3688606 57
CR936825.1 - 14 - 15 1744 57

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC20000264.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EPB41L1

Gene Ontology Categories:

Function(s) Process(es)

cortical actin cytoskeleton organization
cell-cell adhesion
actin binding
structural molecule activity
protein binding
cadherin binding involved in cell-cell adhesion

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR20003635.hg.1 ENST00000202028.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20003635.hg.1 ENST00000338074.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20003635.hg.1 ENST00000344237.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20003635.hg.1 ENST00000373941.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20003635.hg.1 ENST00000373946.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20003635.hg.1 ENST00000373950.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20003635.hg.1 ENST00000373951.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20003635.hg.1 ENST00000397307.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20003635.hg.1 ENST00000397315.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20003635.hg.1 ENST00000441639.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20003635.hg.1 NM_012156.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20003635.hg.1 NM_177996.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20003635.hg.1 TC20000264.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR20003636.hg.1 ENST00000202028.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20003636.hg.1 ENST00000338074.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20003636.hg.1 ENST00000344237.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20003636.hg.1 ENST00000373941.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20003636.hg.1 ENST00000373946.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20003636.hg.1 ENST00000373950.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20003636.hg.1 ENST00000373951.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20003636.hg.1 ENST00000397307.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20003636.hg.1 ENST00000397315.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20003636.hg.1 ENST00000441639.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20003636.hg.1 NM_012156.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20003636.hg.1 NM_177996.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20003636.hg.1 TC20000264.hg.1 HTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0