This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00385505_g1
See other PLD4 GE Assays ›
Gene Symbol:
PLD4
Gene Name:
phospholipase D family member 4
Gene Aliases:
C14orf175
Chromosome Location:
Chr.14: 104924806 - 104937789 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00385505_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PLD4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001308174.1 NP_001295103.1 8 - 9 1270 59
NM_138790.3 NP_620145.2 8 - 9 1232 59
XM_011536411.2 XP_011534713.1 8 - 9 1280 59
XM_017020965.1 XP_016876454.1 8 - 9 1169 59
GenBank mRNA AK295352.1 - 8 - 9 1225 59
AK296188.1 - 8 - 9 1270 59
AY358843.1 - 8 - 9 1198 59
BC015003.1 - 8 - 9 1161 4853026 59

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1400008460.hg.1
This assay targets the same location as HTA transcript cluster(s) TC14000828.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PLD4

Panther Classification:

Molecular Function -

phospholipase

Gene Ontology Categories:

Function(s) Process(es)

lipid catabolic process
inositol phosphate metabolic process
phosphatidylinositol phospholipase C activity
phospholipase D activity
N-acylphosphatidylethanolamine-specific phospholipase D activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR14009056.hg.1 ENST00000392593.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14009056.hg.1 ENST00000540372.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14009056.hg.1 NM_138790.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR14009056.hg.1 TC14000828.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000392593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000392593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000392593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000392593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000392593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000392593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000392593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000392593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000392593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000392593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000392593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000392593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000392593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000392593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000392593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000392593.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000472702.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000472702.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000472702.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000472702.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000472702.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000472901.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000472901.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000472901.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000472901.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000472901.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000540372.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000540372.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000540372.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000540372.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000540372.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000540372.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000540372.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000540372.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000540372.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000540372.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000540372.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000540372.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000540372.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000540372.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000540372.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000540372.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000553861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000553861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000553861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000553861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000553861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000557573.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000557573.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000557573.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000557573.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000557573.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000557573.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 ENST00000557573.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 NM_001308174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 NM_001308174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 NM_001308174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 NM_001308174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 NM_001308174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 NM_001308174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 NM_001308174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 NM_001308174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 NM_001308174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 NM_001308174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 NM_001308174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 NM_001308174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 NM_001308174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 NM_001308174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 NM_001308174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 NM_001308174.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 NM_138790.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 NM_138790.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 NM_138790.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 NM_138790.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 NM_138790.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 NM_138790.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 NM_138790.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 NM_138790.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 NM_138790.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 NM_138790.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 NM_138790.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 NM_138790.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 NM_138790.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 NM_138790.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400008460.hg.1 NM_138790.3 Refseq
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000392593.8 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000392593.8 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000392593.8 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000392593.8 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000392593.8 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000392593.8 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000392593.8 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000392593.8 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000392593.8 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000392593.8 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000392593.8 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000392593.8 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000392593.8 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000392593.8 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000392593.8 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000392593.8 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000472702.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000472702.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000472702.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000472702.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000472702.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000472901.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000472901.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000472901.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000472901.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000472901.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000540372.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000540372.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000540372.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000540372.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000540372.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000540372.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000540372.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000540372.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000540372.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000540372.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000540372.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000540372.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000540372.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000540372.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000540372.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000540372.5 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000553861.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000553861.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000553861.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000553861.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000553861.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000557573.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000557573.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000557573.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000557573.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000557573.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000557573.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 ENST00000557573.1 Ensembl
Affymetrix Human Clariom D Assay TC1400008460.hg.1 NM_001308174.1 Refseq
Affymetrix Human Clariom D Assay TC1400008460.hg.1 NM_001308174.1 Refseq
Affymetrix Human Clariom D Assay TC1400008460.hg.1 NM_001308174.1 Refseq
Affymetrix Human Clariom D Assay TC1400008460.hg.1 NM_001308174.1 Refseq
Affymetrix Human Clariom D Assay TC1400008460.hg.1 NM_001308174.1 Refseq
Affymetrix Human Clariom D Assay TC1400008460.hg.1 NM_001308174.1 Refseq
Affymetrix Human Clariom D Assay TC1400008460.hg.1 NM_001308174.1 Refseq
Affymetrix Human Clariom D Assay TC1400008460.hg.1 NM_001308174.1 Refseq
Affymetrix Human Clariom D Assay TC1400008460.hg.1 NM_001308174.1 Refseq
Affymetrix Human Clariom D Assay TC1400008460.hg.1 NM_001308174.1 Refseq
Affymetrix Human Clariom D Assay TC1400008460.hg.1 NM_001308174.1 Refseq
Affymetrix Human Clariom D Assay TC1400008460.hg.1 NM_001308174.1 Refseq
Affymetrix Human Clariom D Assay TC1400008460.hg.1 NM_001308174.1 Refseq
Affymetrix Human Clariom D Assay TC1400008460.hg.1 NM_001308174.1 Refseq
Affymetrix Human Clariom D Assay TC1400008460.hg.1 NM_001308174.1 Refseq
Affymetrix Human Clariom D Assay TC1400008460.hg.1 NM_001308174.1 Refseq
Affymetrix Human Clariom D Assay TC1400008460.hg.1 NM_138790.3 Refseq
Affymetrix Human Clariom D Assay TC1400008460.hg.1 NM_138790.3 Refseq
Affymetrix Human Clariom D Assay TC1400008460.hg.1 NM_138790.3 Refseq
Affymetrix Human Clariom D Assay TC1400008460.hg.1 NM_138790.3 Refseq
Affymetrix Human Clariom D Assay TC1400008460.hg.1 NM_138790.3 Refseq
Affymetrix Human Clariom D Assay TC1400008460.hg.1 NM_138790.3 Refseq
Affymetrix Human Clariom D Assay TC1400008460.hg.1 NM_138790.3 Refseq
Affymetrix Human Clariom D Assay TC1400008460.hg.1 NM_138790.3 Refseq
Affymetrix Human Clariom D Assay TC1400008460.hg.1 NM_138790.3 Refseq
Affymetrix Human Clariom D Assay TC1400008460.hg.1 NM_138790.3 Refseq
Affymetrix Human Clariom D Assay TC1400008460.hg.1 NM_138790.3 Refseq
Affymetrix Human Clariom D Assay TC1400008460.hg.1 NM_138790.3 Refseq
Affymetrix Human Clariom D Assay TC1400008460.hg.1 NM_138790.3 Refseq
Affymetrix Human Clariom D Assay TC1400008460.hg.1 NM_138790.3 Refseq
Affymetrix Human Clariom D Assay TC1400008460.hg.1 NM_138790.3 Refseq
Affymetrix Human Clariom D Assay PSR1400160635.hg.1 ENST00000392593.8 Ensembl
Affymetrix Human Clariom D Assay PSR1400160635.hg.1 ENST00000472702.1 Ensembl
Affymetrix Human Clariom D Assay PSR1400160635.hg.1 ENST00000540372.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400160635.hg.1 NM_001308174.1 Refseq
Affymetrix Human Clariom D Assay PSR1400160635.hg.1 NM_138790.3 Refseq
Affymetrix Human Clariom D Assay PSR1400160635.hg.1 ENST00000392593.8 Ensembl
Affymetrix Human Clariom D Assay PSR1400160635.hg.1 ENST00000472702.1 Ensembl
Affymetrix Human Clariom D Assay PSR1400160635.hg.1 ENST00000540372.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400160635.hg.1 NM_001308174.1 Refseq
Affymetrix Human Clariom D Assay PSR1400160635.hg.1 NM_138790.3 Refseq
Affymetrix Human Clariom D Assay PSR1400160635.hg.1 TC1400008460.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-r7bw6:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0