This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00385627_m1
See other ORAI1 GE Assays ›
Gene Symbol:
ORAI1
Gene Name:
ORAI calcium release-activated calcium modulator 1
Gene Aliases:
CRACM1, IMD9, ORAT1, TAM2, TMEM142A
Chromosome Location:
Chr.12: 121626550 - 121642040 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00385627_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ORAI1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_032790.3 NP_116179.2 1 - 2 496 110
GenBank mRNA AK027372.1 - 1 - 2 240 110
BC013386.1 - 1 - 2 384 3838524 110
BC015369.1 - 1 - 2 358 3914595 110
BC075831.1 - 1 - 2 496 6645076 110
BC104633.1 - 1 - 2 261 110
BG775018.1 - 1 - 2 223 110
BM762473.1 - 1 - 2 420 110

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200009172.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12000961.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ORAI1

Panther Classification:

Molecular Function -

ion channel transporter

Gene Ontology Categories:

Function(s) Process(es)

store-operated calcium entry
adaptive immune response
regulation of calcium ion transport
positive regulation of calcium ion transport
mammary gland epithelium development
calcium ion import
calcium ion transmembrane transport
protein binding
calmodulin binding
store-operated calcium channel activity
identical protein binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR12012497.hg.1 ENST00000330079.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12012497.hg.1 NM_032790.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12012497.hg.1 TC12000961.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR12012502.hg.1 ENST00000330079.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12012502.hg.1 NM_032790.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12012502.hg.1 TC12000961.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR12012500.hg.1 ENST00000330079.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12012500.hg.1 NM_032790.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12012500.hg.1 TC12000961.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR12012496.hg.1 ENST00000330079.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12012496.hg.1 NM_032790.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12012496.hg.1 TC12000961.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1200009172.hg.1 ENST00000611718.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009172.hg.1 ENST00000611718.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009172.hg.1 ENST00000611718.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009172.hg.1 ENST00000616379.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009172.hg.1 ENST00000616379.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009172.hg.1 ENST00000616379.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009172.hg.1 ENST00000616379.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009172.hg.1 ENST00000616379.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009172.hg.1 ENST00000616379.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009172.hg.1 ENST00000617316.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009172.hg.1 ENST00000617316.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009172.hg.1 ENST00000617316.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009172.hg.1 ENST00000617316.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009172.hg.1 ENST00000617316.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009172.hg.1 ENST00000617316.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009172.hg.1 ENST00000617316.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200009172.hg.1 NM_032790.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009172.hg.1 NM_032790.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009172.hg.1 NM_032790.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009172.hg.1 NM_032790.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009172.hg.1 NM_032790.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200009172.hg.1 NM_032790.3 Refseq
Affymetrix Human Clariom D Assay PSR1200167863.hg.1 ENST00000616379.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200167863.hg.1 ENST00000617316.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200167863.hg.1 NM_032790.3 Refseq
Affymetrix Human Clariom D Assay PSR1200167863.hg.1 ENST00000616379.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200167863.hg.1 ENST00000617316.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200167863.hg.1 NM_032790.3 Refseq
Affymetrix Human Clariom D Assay PSR1200167863.hg.1 TC1200009172.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1200167864.hg.1 ENST00000616379.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200167864.hg.1 ENST00000617316.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200167864.hg.1 NM_032790.3 Refseq
Affymetrix Human Clariom D Assay PSR1200167864.hg.1 ENST00000616379.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200167864.hg.1 ENST00000617316.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200167864.hg.1 NM_032790.3 Refseq
Affymetrix Human Clariom D Assay PSR1200167864.hg.1 TC1200009172.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1200167866.hg.1 ENST00000611718.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200167866.hg.1 ENST00000616379.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200167866.hg.1 ENST00000617316.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200167866.hg.1 NM_032790.3 Refseq
Affymetrix Human Clariom D Assay PSR1200167866.hg.1 ENST00000611718.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200167866.hg.1 ENST00000616379.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200167866.hg.1 ENST00000617316.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200167866.hg.1 NM_032790.3 Refseq
Affymetrix Human Clariom D Assay PSR1200167866.hg.1 TC1200009172.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1200009172.hg.1 ENST00000611718.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009172.hg.1 ENST00000611718.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009172.hg.1 ENST00000611718.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009172.hg.1 ENST00000616379.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009172.hg.1 ENST00000616379.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009172.hg.1 ENST00000616379.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009172.hg.1 ENST00000616379.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009172.hg.1 ENST00000616379.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009172.hg.1 ENST00000616379.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009172.hg.1 ENST00000617316.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009172.hg.1 ENST00000617316.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009172.hg.1 ENST00000617316.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009172.hg.1 ENST00000617316.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009172.hg.1 ENST00000617316.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009172.hg.1 ENST00000617316.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009172.hg.1 ENST00000617316.1 Ensembl
Affymetrix Human Clariom D Assay TC1200009172.hg.1 NM_032790.3 Refseq
Affymetrix Human Clariom D Assay TC1200009172.hg.1 NM_032790.3 Refseq
Affymetrix Human Clariom D Assay TC1200009172.hg.1 NM_032790.3 Refseq
Affymetrix Human Clariom D Assay TC1200009172.hg.1 NM_032790.3 Refseq
Affymetrix Human Clariom D Assay TC1200009172.hg.1 NM_032790.3 Refseq
Affymetrix Human Clariom D Assay TC1200009172.hg.1 NM_032790.3 Refseq
Affymetrix Human Clariom D Assay PSR1200167865.hg.1 ENST00000616379.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200167865.hg.1 ENST00000617316.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200167865.hg.1 NM_032790.3 Refseq
Affymetrix Human Clariom D Assay PSR1200167865.hg.1 ENST00000616379.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200167865.hg.1 ENST00000617316.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200167865.hg.1 NM_032790.3 Refseq
Affymetrix Human Clariom D Assay PSR1200167865.hg.1 TC1200009172.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 144226 - -
ABI Human Genome Survey Microarray Version 2.0 199779 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0