This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00385748_m1
See other PPP1R14C GE Assays ›
Gene Symbol:
PPP1R14C
Gene Name:
protein phosphatase 1 regulatory inhibitor subunit 14C
Gene Aliases:
CPI17-like, KEPI, NY-BR-81
Chromosome Location:
Chr.6: 150143052 - 150250392 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00385748_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPP1R14C
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_030949.2 NP_112211.1 1 - 2 441 118
GenBank mRNA AF308297.1 - 1 - 2 314 118
AF407165.2 - 1 - 2 943 118
AK096272.1 - 1 - 2 77 118
BC015773.1 - 1 - 2 417 4877511 118
CB994599.1 - 1 - 2 469 118
DR003369.1 - 1 - 2 451 118

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600009831.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06001091.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PPP1R14C

Panther Classification:

Molecular Function -

phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

regulation of protein dephosphorylation
regulation of phosphorylation
negative regulation of catalytic activity
protein serine/threonine phosphatase inhibitor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR06012876.hg.1 TC06001091.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR06012876.hg.1 ENST00000361131.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06012876.hg.1 NM_030949.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06012875.hg.1 TC06001091.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR06012875.hg.1 ENST00000361131.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06012875.hg.1 NM_030949.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009831.hg.1 ENST00000361131.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009831.hg.1 ENST00000361131.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009831.hg.1 ENST00000361131.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009831.hg.1 ENST00000361131.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009831.hg.1 ENST00000361131.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009831.hg.1 ENST00000361131.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009831.hg.1 ENST00000361131.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009831.hg.1 NM_030949.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009831.hg.1 NM_030949.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009831.hg.1 NM_030949.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009831.hg.1 NM_030949.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009831.hg.1 NM_030949.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009831.hg.1 NM_030949.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009831.hg.1 NM_030949.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009831.hg.1 NM_030949.2 Refseq
Affymetrix Human Clariom D Assay PSR0600167275.hg.1 ENST00000361131.4 Ensembl
Affymetrix Human Clariom D Assay PSR0600167275.hg.1 NM_030949.2 Refseq
Affymetrix Human Clariom D Assay PSR0600167275.hg.1 ENST00000361131.4 Ensembl
Affymetrix Human Clariom D Assay PSR0600167275.hg.1 NM_030949.2 Refseq
Affymetrix Human Clariom D Assay PSR0600167275.hg.1 TC0600009831.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0600167274.hg.1 ENST00000361131.4 Ensembl
Affymetrix Human Clariom D Assay PSR0600167274.hg.1 NM_030949.2 Refseq
Affymetrix Human Clariom D Assay PSR0600167274.hg.1 ENST00000361131.4 Ensembl
Affymetrix Human Clariom D Assay PSR0600167274.hg.1 NM_030949.2 Refseq
Affymetrix Human Clariom D Assay PSR0600167274.hg.1 TC0600009831.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0600009831.hg.1 ENST00000361131.4 Ensembl
Affymetrix Human Clariom D Assay TC0600009831.hg.1 ENST00000361131.4 Ensembl
Affymetrix Human Clariom D Assay TC0600009831.hg.1 ENST00000361131.4 Ensembl
Affymetrix Human Clariom D Assay TC0600009831.hg.1 ENST00000361131.4 Ensembl
Affymetrix Human Clariom D Assay TC0600009831.hg.1 ENST00000361131.4 Ensembl
Affymetrix Human Clariom D Assay TC0600009831.hg.1 ENST00000361131.4 Ensembl
Affymetrix Human Clariom D Assay TC0600009831.hg.1 ENST00000361131.4 Ensembl
Affymetrix Human Clariom D Assay TC0600009831.hg.1 NM_030949.2 Refseq
Affymetrix Human Clariom D Assay TC0600009831.hg.1 NM_030949.2 Refseq
Affymetrix Human Clariom D Assay TC0600009831.hg.1 NM_030949.2 Refseq
Affymetrix Human Clariom D Assay TC0600009831.hg.1 NM_030949.2 Refseq
Affymetrix Human Clariom D Assay TC0600009831.hg.1 NM_030949.2 Refseq
Affymetrix Human Clariom D Assay TC0600009831.hg.1 NM_030949.2 Refseq
Affymetrix Human Clariom D Assay TC0600009831.hg.1 NM_030949.2 Refseq
Affymetrix Human Clariom D Assay TC0600009831.hg.1 NM_030949.2 Refseq
Affymetrix Human Clariom D Assay PSR0600167273.hg.1 ENST00000361131.4 Ensembl
Affymetrix Human Clariom D Assay PSR0600167273.hg.1 NM_030949.2 Refseq
Affymetrix Human Clariom D Assay PSR0600167273.hg.1 ENST00000361131.4 Ensembl
Affymetrix Human Clariom D Assay PSR0600167273.hg.1 NM_030949.2 Refseq
Affymetrix Human Clariom D Assay PSR0600167273.hg.1 TC0600009831.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-s2cfr:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0