This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00388675_m1
See other TAP1 GE Assays ›
Gene Symbol:
TAP1
Gene Name:
transporter 1, ATP binding cassette subfamily B member
Gene Aliases:
ABC17, ABCB2, APT1, D6S114E, PSF-1, PSF1, RING4, TAP1*0102N, TAP1N
Chromosome Location:
Chr.6: 32845209 - 32853971 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00388675_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TAP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000593.5 NP_000584.2 3 - 4 1177 74
NM_001292022.1 NP_001278951.1 3 - 4 437 74
GenBank mRNA AB012644.1 - 3 - 4 841 74
AK302340.1 - 3 - 4 408 74
AK313762.1 - 3 - 4 1213 74
AY523970.1 - 3 - 4 1048 74
AY523971.2 - 3 - 4 1048 74
BC014081.2 - 3 - 4 1177 4304716 74
BX648013.1 - 3 - 4 1185 74
DC354917.1 - 3 - 4 437 74
KU178346.1 - 3 - 4 1022 74
L21204.1 - 3 - 4 842 74
L21205.1 - 3 - 4 842 74
L21206.1 - 3 - 4 842 74
L21207.1 - 3 - 4 842 74
L21208.1 - 3 - 4 842 74
X57521.1 - 3 - 4 476 74
X57522.1 - 3 - 4 1052 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600011508.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06001574.hg.1, TC6_apd_hap1000105.hg.1, TC6_cox_hap2000199.hg.1, TC6_dbb_hap3000185.hg.1, TC6_mann_hap4000167.hg.1, TC6_mcf_hap5000174.hg.1, TC6_qbl_hap6000187.hg.1, TC6_ssto_hap7000169.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Recommended Assay Homo sapiens Human Drug Transporters 1700 Inventoried Human_Drug_Transporters Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TAP1

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter

Gene Ontology Categories:

Function(s) Process(es)

adaptive immune response
antigen processing and presentation of peptide antigen via MHC class I
antigen processing and presentation of exogenous protein antigen via MHC class Ib, TAP-dependent
antigen processing and presentation of endogenous peptide antigen via MHC class I via ER pathway, TAP-dependent
antigen processing and presentation of endogenous peptide antigen via MHC class Ib via ER pathway, TAP-dependent
positive regulation of antigen processing and presentation of peptide antigen via MHC class I
defense response
peptide transport
intracellular transport of viral protein in host cell
antigen processing and presentation of endogenous peptide antigen via MHC class I
protection from natural killer cell mediated cytotoxicity
cytosol to ER transport
transmembrane transport
protein binding
ATP binding
peptide transporter activity
peptide-transporting ATPase activity
MHC class Ib protein binding
MHC class I protein binding
peptide antigen binding
ATPase activity, coupled to transmembrane movement of substances
protein homodimerization activity
ADP binding
TAP1 binding
TAP2 binding
tapasin binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000354258.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000354258.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000354258.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000354258.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000354258.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000354258.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000354258.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000354258.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000354258.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000354258.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000354258.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000354258.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000354258.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000354258.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000486332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000486332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000486332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000486332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000486332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000486332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000486332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000486332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000486332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000486332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000487296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000487296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000487296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000487296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000487296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000487296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 NM_001292022.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 NM_001292022.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 NM_001292022.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 NM_001292022.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 NM_001292022.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 NM_001292022.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 NM_001292022.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 NM_001292022.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 NM_001292022.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 NM_001292022.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 NM_001292022.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 NM_001292022.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 NM_001292022.1 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 151778 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline