This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00388932_m1
See other MGST3 GE Assays ›
Gene Symbol:
MGST3
Gene Name:
microsomal glutathione S-transferase 3
Gene Aliases:
GST-III
Chromosome Location:
Chr.1: 165630873 - 165656136 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00388932_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MGST3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_004528.3 NP_004519.1 4 - 5 680 94
XM_005245174.3 XP_005245231.2 4 - 5 283 94
GenBank mRNA AF026977.1 - 3 - 4 301 94
AK223012.1 - 3 - 4 280 94
AK312103.1 - 4 - 5 296 94
BC000505.2 - 4 - 5 279 2822075 94
BC003034.2 - 3 - 4 261 2822075 94
BC005964.1 - 4 - 5 329 4068898 94
BI666647.1 - 4 - 5 345 94
CR450359.1 - 3 - 4 252 94

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100010454.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01001443.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MGST3

Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
xenobiotic metabolic process
signal transduction
response to organonitrogen compound
oxidation-reduction process
cellular oxidant detoxification
glutathione derivative biosynthetic process
glutathione transferase activity
peroxidase activity
glutathione peroxidase activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01022490.hg.1 ENST00000367883.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01022490.hg.1 ENST00000367884.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01022490.hg.1 ENST00000367885.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01022490.hg.1 ENST00000367886.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01022490.hg.1 ENST00000367888.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01022490.hg.1 ENST00000367889.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01022490.hg.1 ENST00000404549.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01022490.hg.1 NM_004528.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01022490.hg.1 TC01001443.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01022493.hg.1 ENST00000367883.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01022493.hg.1 ENST00000367884.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01022493.hg.1 ENST00000367885.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01022493.hg.1 ENST00000367886.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01022493.hg.1 ENST00000367888.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01022493.hg.1 ENST00000367889.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01022493.hg.1 ENST00000404549.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01022493.hg.1 NM_004528.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01022493.hg.1 TC01001443.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000367883.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000367883.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000367883.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000367883.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000367883.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000367883.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000367883.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000367883.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000367884.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000367884.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000367884.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000367884.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000367884.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000367884.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000367884.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000367885.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000367885.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000367885.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000367885.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000367885.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000367885.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000367885.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000367888.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000367888.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000367888.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000367888.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000367888.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000367889.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000367889.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000367889.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000367889.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000367889.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000367889.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000367889.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000460632.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000461308.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000461308.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000477450.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000477450.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000477450.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000477450.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000488688.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000488688.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000488688.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000494074.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000494074.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000494074.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000494074.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000495447.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000495447.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000495447.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000495447.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000495447.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000495447.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000495447.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000609263.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000609263.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000609263.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000609263.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000609263.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000609263.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000609263.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000609263.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000627653.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000627653.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000627653.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000627653.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000627653.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000627653.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000627653.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 ENST00000627653.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 NM_004528.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 NM_004528.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 NM_004528.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 NM_004528.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 NM_004528.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 NM_004528.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 NM_004528.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100010454.hg.1 NM_004528.3 Refseq
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000367883.3 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000367883.3 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000367883.3 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000367883.3 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000367883.3 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000367883.3 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000367883.3 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000367883.3 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000367884.6 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000367884.6 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000367884.6 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000367884.6 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000367884.6 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000367884.6 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000367884.6 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000367885.5 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000367885.5 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000367885.5 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000367885.5 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000367885.5 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000367885.5 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000367885.5 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000367888.8 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000367888.8 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000367888.8 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000367888.8 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000367888.8 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000367889.7 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000367889.7 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000367889.7 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000367889.7 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000367889.7 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000367889.7 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000367889.7 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000460632.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000461308.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000461308.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000477450.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000477450.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000477450.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000477450.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000488688.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000488688.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000488688.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000494074.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000494074.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000494074.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000494074.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000495447.5 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000495447.5 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000495447.5 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000495447.5 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000495447.5 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000495447.5 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000495447.5 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000609263.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000609263.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000609263.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000609263.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000609263.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000609263.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000609263.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000609263.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000627653.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000627653.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000627653.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000627653.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000627653.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000627653.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000627653.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 ENST00000627653.1 Ensembl
Affymetrix Human Clariom D Assay TC0100010454.hg.1 NM_004528.3 Refseq
Affymetrix Human Clariom D Assay TC0100010454.hg.1 NM_004528.3 Refseq
Affymetrix Human Clariom D Assay TC0100010454.hg.1 NM_004528.3 Refseq
Affymetrix Human Clariom D Assay TC0100010454.hg.1 NM_004528.3 Refseq
Affymetrix Human Clariom D Assay TC0100010454.hg.1 NM_004528.3 Refseq
Affymetrix Human Clariom D Assay TC0100010454.hg.1 NM_004528.3 Refseq
Affymetrix Human Clariom D Assay TC0100010454.hg.1 NM_004528.3 Refseq
Affymetrix Human Clariom D Assay TC0100010454.hg.1 NM_004528.3 Refseq
Affymetrix Human Clariom D Assay PSR0100176962.hg.1 ENST00000367883.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100176962.hg.1 ENST00000367884.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100176962.hg.1 ENST00000367885.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100176962.hg.1 ENST00000367888.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100176962.hg.1 ENST00000367889.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100176962.hg.1 ENST00000494074.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100176962.hg.1 ENST00000495447.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100176962.hg.1 ENST00000627653.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100176962.hg.1 NM_004528.3 Refseq
Affymetrix Human Clariom D Assay PSR0100176962.hg.1 ENST00000367883.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100176962.hg.1 ENST00000367884.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100176962.hg.1 ENST00000367885.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100176962.hg.1 ENST00000367888.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100176962.hg.1 ENST00000367889.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100176962.hg.1 ENST00000494074.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100176962.hg.1 ENST00000495447.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100176962.hg.1 ENST00000627653.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100176962.hg.1 NM_004528.3 Refseq
Affymetrix Human Clariom D Assay PSR0100176962.hg.1 TC0100010454.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0100176959.hg.1 ENST00000367883.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100176959.hg.1 ENST00000367884.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100176959.hg.1 ENST00000367885.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100176959.hg.1 ENST00000367888.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100176959.hg.1 ENST00000367889.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100176959.hg.1 ENST00000477450.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100176959.hg.1 ENST00000495447.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100176959.hg.1 ENST00000627653.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100176959.hg.1 NM_004528.3 Refseq
Affymetrix Human Clariom D Assay PSR0100176959.hg.1 ENST00000367883.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100176959.hg.1 ENST00000367884.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100176959.hg.1 ENST00000367885.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100176959.hg.1 ENST00000367888.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100176959.hg.1 ENST00000367889.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100176959.hg.1 ENST00000477450.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100176959.hg.1 ENST00000495447.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100176959.hg.1 ENST00000627653.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100176959.hg.1 NM_004528.3 Refseq
Affymetrix Human Clariom D Assay PSR0100176959.hg.1 TC0100010454.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline