This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00389130_m1
See other RAMP3 GE Assays ›
Gene Symbol:
RAMP3
Gene Name:
receptor (G protein-coupled) activity modifying protein 3
Gene Aliases:
-
Chromosome Location:
Chr.7: 45157768 - 45184250 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00389130_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RAMP3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005856.2 NP_005847.1 1 - 2 118 94
GenBank mRNA AJ001016.1 - 1 - 2 86 94
AK311871.1 - 1 - 2 57 94
AY265459.1 - 1 - 2 57 94
BC022304.1 - 1 - 2 85 4717934 94
BC053852.1 - 1 - 2 84 6197363 94
BP347072.1 - 1 - 2 118 94

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RAMP3

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of receptor recycling
calcium ion transport
intracellular protein transport
regulation of G-protein coupled receptor protein signaling pathway
protein transport
receptor internalization
negative regulation of transcription, DNA-templated
cellular response to estradiol stimulus
protein localization to plasma membrane
G-protein coupled receptor signaling pathway involved in heart process
positive regulation of establishment of protein localization to plasma membrane
receptor activity
protein binding
protein transporter activity
coreceptor activity

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 122061 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0