This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00393468_m1
See other PPP1R9A GE Assays ›
Gene Symbol:
PPP1R9A
Gene Name:
protein phosphatase 1 regulatory subunit 9A
Gene Aliases:
NRB1, NRBI, Neurabin-I
Chromosome Location:
Chr.7: 94907202 - 95296415 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00393468_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPP1R9A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017650.2 NP_060120.2 14 - 15 3177 90
XM_017012395.1 XP_016867884.1 20 - 21 3978 90
XM_017012396.1 XP_016867885.1 18 - 19 4087 90
XM_017012399.1 XP_016867888.1 18 - 19 3909 90
XM_017012400.1 XP_016867889.1 18 - 19 3849 90
XM_017012402.1 XP_016867891.1 18 - 19 3795 90
XM_017012404.1 XP_016867893.1 16 - 17 3306 90
XM_017012405.1 XP_016867894.1 14 - 15 3301 90
XM_017012406.1 XP_016867895.1 14 - 15 3232 90
GenBank mRNA AB033048.1 - 13 - 14 1829 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700008375.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07000565.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PPP1R9A

Gene Ontology Categories:

Function(s) Process(es)

actin filament organization
aging
calcium-mediated signaling
regulation of actin filament polymerization
neuron projection development
positive regulation of protein kinase activity
regulation of filopodium assembly
negative regulation of stress fiber assembly
regulation of synapse structural plasticity
regulation of synapse assembly
excitatory postsynaptic potential
positive regulation of dendritic spine development
regulation of dendritic spine morphogenesis
cellular response to toxic substance
negative regulation of long-term synaptic potentiation
positive regulation of long term synaptic depression
negative regulation of spontaneous neurotransmitter secretion
protein binding
protein C-terminus binding
protein phosphatase 1 binding
protein kinase binding
protein domain specific binding
protein homodimerization activity
ion channel binding
actin filament binding
GTPase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR07008348.hg.1 ENST00000289495.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07008348.hg.1 ENST00000340694.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07008348.hg.1 ENST00000433360.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07008348.hg.1 ENST00000433881.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07008348.hg.1 NM_001166160.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07008348.hg.1 NM_001166161.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07008348.hg.1 NM_001166163.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07008348.hg.1 NM_017650.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07008348.hg.1 TC07000565.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0700158560.hg.1 ENST00000289495.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700158560.hg.1 ENST00000340694.8 Ensembl
Affymetrix Human Clariom D Assay PSR0700158560.hg.1 ENST00000433360.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700158560.hg.1 ENST00000433881.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700158560.hg.1 NM_001166160.1 Refseq
Affymetrix Human Clariom D Assay PSR0700158560.hg.1 NM_001166161.1 Refseq
Affymetrix Human Clariom D Assay PSR0700158560.hg.1 NM_001166163.1 Refseq
Affymetrix Human Clariom D Assay PSR0700158560.hg.1 NM_017650.2 Refseq
Affymetrix Human Clariom D Assay PSR0700158560.hg.1 ENST00000289495.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700158560.hg.1 ENST00000340694.8 Ensembl
Affymetrix Human Clariom D Assay PSR0700158560.hg.1 ENST00000433360.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700158560.hg.1 ENST00000433881.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700158560.hg.1 NM_001166160.1 Refseq
Affymetrix Human Clariom D Assay PSR0700158560.hg.1 NM_001166161.1 Refseq
Affymetrix Human Clariom D Assay PSR0700158560.hg.1 NM_001166163.1 Refseq
Affymetrix Human Clariom D Assay PSR0700158560.hg.1 NM_017650.2 Refseq
Affymetrix Human Clariom D Assay PSR0700158560.hg.1 TC0700008375.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline