This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00393705_m1
See other SGPL1 GE Assays ›
Gene Symbol:
SGPL1
Gene Name:
sphingosine-1-phosphate lyase 1
Gene Aliases:
S1PL, SPL
Chromosome Location:
Chr.10: 70815920 - 70881189 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00393705_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SGPL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003901.3 NP_003892.2 7 - 8 837 83
XM_005270263.1 XP_005270320.1 7 - 8 850 83
XM_006718053.1 XP_006718116.1 8 - 9 952 83
XM_011540316.2 XP_011538618.1 7 - 8 896 83
XM_011540317.1 XP_011538619.1 7 - 8 746 83
XM_011540318.1 XP_011538620.1 6 - 7 767 83
GenBank mRNA AB033078.1 - 7 - 8 815 83
AJ011304.2 - 7 - 8 792 83
AK314615.1 - 7 - 8 746 83
BC052991.1 - 7 - 8 825 6150776 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000007936.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10000434.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SGPL1

Gene Ontology Categories:

Function(s) Process(es)

luteinization
vasculogenesis
kidney development
fatty acid metabolic process
ceramide metabolic process
spermatogenesis
androgen metabolic process
estrogen metabolic process
post-embryonic development
fibroblast migration
hemopoiesis
sphingolipid biosynthetic process
sphingolipid catabolic process
Leydig cell differentiation
regulation of multicellular organism growth
platelet-derived growth factor receptor signaling pathway
skeletal system morphogenesis
palate development
face morphogenesis
apoptotic signaling pathway
protein binding
sphinganine-1-phosphate aldolase activity
carboxy-lyase activity
pyridoxal phosphate binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR10005097.hg.1 ENST00000373202.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10005097.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10005097.hg.1 TC10000434.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR10005094.hg.1 ENST00000373202.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10005094.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10005094.hg.1 TC10000434.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000299297.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000299297.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000299297.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000299297.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000299297.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000299297.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000409118.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000409118.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000409118.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000409118.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000486993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000486993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000486993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000486993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000611290.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000611290.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000611290.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000611290.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000613857.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000613857.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000613857.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000613857.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000613857.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000613857.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000618021.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000618021.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000618021.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000618021.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000618021.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000618021.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000620724.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000620724.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000620724.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000620724.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000620724.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000620724.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 ENST00000620724.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay PSR1000153876.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000153876.hg.1 ENST00000613857.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000153876.hg.1 ENST00000620724.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000153876.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay PSR1000153876.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000153876.hg.1 ENST00000613857.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000153876.hg.1 ENST00000620724.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000153876.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay PSR1000153876.hg.1 TC1000007936.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000299297.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000299297.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000299297.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000299297.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000299297.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000299297.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000409118.2 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000409118.2 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000409118.2 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000409118.2 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000486993.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000486993.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000486993.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000486993.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000611290.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000611290.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000611290.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000611290.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000613857.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000613857.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000613857.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000613857.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000613857.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000613857.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000618021.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000618021.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000618021.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000618021.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000618021.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000618021.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000620724.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000620724.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000620724.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000620724.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000620724.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000620724.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 ENST00000620724.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay TC1000007936.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay PSR1000153871.hg.1 ENST00000299297.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000153871.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000153871.hg.1 ENST00000409118.2 Ensembl
Affymetrix Human Clariom D Assay PSR1000153871.hg.1 ENST00000611290.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000153871.hg.1 ENST00000613857.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000153871.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay PSR1000153871.hg.1 ENST00000299297.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000153871.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000153871.hg.1 ENST00000409118.2 Ensembl
Affymetrix Human Clariom D Assay PSR1000153871.hg.1 ENST00000611290.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000153871.hg.1 ENST00000613857.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000153871.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay PSR1000153871.hg.1 TC1000007936.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1000153875.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000153875.hg.1 ENST00000611290.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000153875.hg.1 ENST00000613857.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000153875.hg.1 ENST00000620724.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000153875.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay PSR1000153875.hg.1 ENST00000373202.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000153875.hg.1 ENST00000611290.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000153875.hg.1 ENST00000613857.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000153875.hg.1 ENST00000620724.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000153875.hg.1 NM_003901.3 Refseq
Affymetrix Human Clariom D Assay PSR1000153875.hg.1 TC1000007936.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0