| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04023257.hg.1 |
ENST00000537611.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04023257.hg.1 |
NM_017419.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04023257.hg.1 |
TC04001669.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
ENST00000537611.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400012212.hg.1 |
NM_017419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400183870.hg.1 |
ENST00000537611.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400183870.hg.1 |
NM_017419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400183870.hg.1 |
ENST00000537611.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400183870.hg.1 |
NM_017419.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400183870.hg.1 |
TC0400012212.hg.1 |
Clariom D TC |