This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00398903_m1
See other GLT8D2 GE Assays ›
Gene Symbol:
GLT8D2
Gene Name:
glycosyltransferase 8 domain containing 2
Gene Aliases:
-
Chromosome Location:
Chr.12: 103988983 - 104064177 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00398903_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GLT8D2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001316967.1 NP_001303896.1 7 - 8 1142 86
NM_031302.4 NP_112592.1 8 - 9 1277 86
XM_011538793.2 XP_011537095.1 8 - 9 1512 86
XM_017019999.1 XP_016875488.1 9 - 10 1593 86
GenBank mRNA AJ271711.1 - 5 - 6 491 86
AK027310.1 - 6 - 7 591 86
AK056056.1 - 7 - 8 1148 86
AY358763.1 - 7 - 8 681 86
BC022343.1 - 7 - 8 898 4248949 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200011752.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12001904.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GLT8D2

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s) Process(es)

polysaccharide biosynthetic process
transferase activity, transferring glycosyl groups

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR12024846.hg.1 ENST00000360814.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12024846.hg.1 ENST00000546436.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12024846.hg.1 ENST00000548660.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12024846.hg.1 NM_031302.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12024846.hg.1 TC12001904.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000360814.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000360814.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000360814.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000360814.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000360814.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000360814.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000360814.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000360814.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000360814.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000360814.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000360814.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000360814.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000360814.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000360814.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000546436.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000546436.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000546436.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000546436.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000546436.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000546436.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000546436.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000546436.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000546436.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000546436.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000546436.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000546436.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000546851.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000546851.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000546851.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000546851.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000546851.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000547583.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000547583.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000547583.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000547583.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000547583.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000547583.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000547583.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000548660.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000548660.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000548660.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000548660.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000548660.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000548660.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000548660.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000548660.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000548660.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000548660.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000548660.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000548660.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000548660.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000548660.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000550816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000550816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000550816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000550816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000550816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000550816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000552572.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000552572.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 ENST00000552572.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 NM_031302.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 NM_031302.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 NM_031302.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 NM_031302.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 NM_031302.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 NM_031302.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 NM_031302.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 NM_031302.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 NM_031302.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 NM_031302.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 NM_031302.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 NM_031302.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 NM_031302.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011752.hg.1 NM_031302.4 Refseq
Affymetrix Human Clariom D Assay PSR1200191132.hg.1 ENST00000360814.8 Ensembl
Affymetrix Human Clariom D Assay PSR1200191132.hg.1 ENST00000546436.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200191132.hg.1 ENST00000548660.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200191132.hg.1 NM_031302.4 Refseq
Affymetrix Human Clariom D Assay PSR1200191132.hg.1 ENST00000360814.8 Ensembl
Affymetrix Human Clariom D Assay PSR1200191132.hg.1 ENST00000546436.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200191132.hg.1 ENST00000548660.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200191132.hg.1 NM_031302.4 Refseq
Affymetrix Human Clariom D Assay PSR1200191132.hg.1 TC1200011752.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000360814.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000360814.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000360814.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000360814.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000360814.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000360814.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000360814.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000360814.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000360814.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000360814.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000360814.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000360814.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000360814.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000360814.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000546436.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000546436.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000546436.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000546436.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000546436.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000546436.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000546436.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000546436.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000546436.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000546436.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000546436.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000546436.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000546851.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000546851.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000546851.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000546851.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000546851.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000547583.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000547583.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000547583.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000547583.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000547583.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000547583.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000547583.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000548660.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000548660.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000548660.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000548660.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000548660.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000548660.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000548660.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000548660.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000548660.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000548660.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000548660.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000548660.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000548660.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000548660.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000550816.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000550816.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000550816.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000550816.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000550816.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000550816.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000552572.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000552572.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 ENST00000552572.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011752.hg.1 NM_031302.4 Refseq
Affymetrix Human Clariom D Assay TC1200011752.hg.1 NM_031302.4 Refseq
Affymetrix Human Clariom D Assay TC1200011752.hg.1 NM_031302.4 Refseq
Affymetrix Human Clariom D Assay TC1200011752.hg.1 NM_031302.4 Refseq
Affymetrix Human Clariom D Assay TC1200011752.hg.1 NM_031302.4 Refseq
Affymetrix Human Clariom D Assay TC1200011752.hg.1 NM_031302.4 Refseq
Affymetrix Human Clariom D Assay TC1200011752.hg.1 NM_031302.4 Refseq
Affymetrix Human Clariom D Assay TC1200011752.hg.1 NM_031302.4 Refseq
Affymetrix Human Clariom D Assay TC1200011752.hg.1 NM_031302.4 Refseq
Affymetrix Human Clariom D Assay TC1200011752.hg.1 NM_031302.4 Refseq
Affymetrix Human Clariom D Assay TC1200011752.hg.1 NM_031302.4 Refseq
Affymetrix Human Clariom D Assay TC1200011752.hg.1 NM_031302.4 Refseq
Affymetrix Human Clariom D Assay TC1200011752.hg.1 NM_031302.4 Refseq
Affymetrix Human Clariom D Assay TC1200011752.hg.1 NM_031302.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0