This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00399785_m1
See other SUMF1 GE Assays ›
Gene Symbol:
SUMF1
Gene Name:
sulfatase modifying factor 1
Gene Aliases:
AAPA3037, FGE, UNQ3037
Chromosome Location:
Chr.3: 4034714 - 4467282 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00399785_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SUMF1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001164674.1 NP_001158146.1 5 - 6 802 83
NM_001164675.1 NP_001158147.1 6 - 7 877 83
NM_182760.3 NP_877437.2 6 - 7 877 83
XM_011533624.2 XP_011531926.1 6 - 7 868 83
XM_011533625.2 XP_011531927.1 6 - 7 868 83
XM_011533626.2 XP_011531928.1 6 - 7 868 83
XM_017006252.1 XP_016861741.1 6 - 7 868 83
XM_017006253.1 XP_016861742.1 5 - 6 790 83
XM_017006254.1 XP_016861743.1 6 - 7 868 83
XM_017006255.1 XP_016861744.1 6 - 7 868 83
GenBank mRNA AB448737.1 - 6 - 7 848 83
AK057983.1 - 7 - 8 776 83
AK075459.1 - 6 - 7 859 83
AK302018.1 - 5 - 6 770 83
AY208752.1 - 6 - 7 840 83
AY323910.1 - 6 - 7 840 83
AY358092.1 - 6 - 7 857 83
BC017005.2 - 1 - 2 116 83
BC110862.1 - 6 - 7 848 5760490 83
BC121122.2 - 6 - 7 869 40120670 83
BC121123.2 - 6 - 7 869 40120671 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300013926.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03001125.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SUMF1

Gene Ontology Categories:

Function(s) Process(es)

glycosphingolipid metabolic process
post-translational protein modification
oxidation-reduction process
oxidoreductase activity
protein homodimerization activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000272902.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000383843.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000383843.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000383843.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000383843.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000383843.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000383843.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000383843.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000383843.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000383843.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000383843.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000383843.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000405420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000405420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000405420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000405420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000405420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000405420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000405420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000405420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000405420.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000431226.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000458465.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000458465.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000458465.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000458465.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000458465.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000458465.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 ENST00000458465.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_001164675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013926.hg.1 NM_182760.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0