Assay Details
| Gene Symbol: PRSS54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080492.1 | NP_001073961.1 | 3 - 4 | 486 | 72 | |
| NM_001305173.1 | NP_001292102.1 | 3 - 4 | 483 | 72 | ||
| GenBank mRNA | AK058068.1 | - | 3 - 4 | 428 | 72 | |
| BC057843.1 | - | 3 - 4 | 475 | 5296090 | 72 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001151.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16016024.hg.1 | ENST00000219301.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016024.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16016024.hg.1 | TC16001151.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000563336.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000563336.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000563336.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000563336.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000563336.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000563336.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000563336.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000563336.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180592.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180592.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180592.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180592.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180592.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180592.hg.1 | TC1600010480.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600180592.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180592.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180592.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180592.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180592.hg.1 | NM_001305173.1 | Refseq |

PRSS54
Genomic Map