Assay Details
| Gene Symbol: CLNK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_052964.2 | NP_443196.2 | 7 - 8 | 540 | 62 | |
| XM_011513775.2 | XP_011512077.1 | 7 - 8 | 771 | 62 | ||
| XM_017007684.1 | XP_016863173.1 | 7 - 8 | 771 | 62 | ||
| XM_017007685.1 | XP_016863174.1 | 7 - 8 | 771 | 62 | ||
| GenBank mRNA | AK093001.1 | - | 7 - 8 | 542 | 62 | |
| BC029887.1 | - | 7 - 8 | 570 | 62 | ||
| BC137553.1 | - | 7 - 8 | 469 | 9021553 | 62 | |
| BC137556.1 | - | 7 - 8 | 469 | 9021556 | 62 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001036.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04014233.hg.1 | ENST00000226951.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014233.hg.1 | ENST00000429087.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014233.hg.1 | ENST00000442825.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014233.hg.1 | ENST00000507719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014233.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014233.hg.1 | TC04001036.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000507719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000507719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000507719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000507719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000507719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000507719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000507719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000507719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000507719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000507719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000507719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000507719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000515667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000515667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000515667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000515667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000515667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000515667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000515667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | ENST00000515667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014232.hg.1 | ENST00000226951.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014232.hg.1 | ENST00000429087.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014232.hg.1 | ENST00000442825.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014232.hg.1 | ENST00000507719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014232.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04014232.hg.1 | TC04001036.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0400168927.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400168927.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400168927.hg.1 | ENST00000507719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400168927.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400168927.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400168927.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400168927.hg.1 | ENST00000507719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400168927.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400168927.hg.1 | TC0400010050.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000507719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000507719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000507719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000507719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000507719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000507719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000507719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000507719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000507719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000507719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000507719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000507719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000515667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000515667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000515667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000515667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000515667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000515667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000515667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | ENST00000515667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010050.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400168928.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400168928.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400168928.hg.1 | ENST00000507719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400168928.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400168928.hg.1 | ENST00000226951.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400168928.hg.1 | ENST00000442825.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400168928.hg.1 | ENST00000507719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400168928.hg.1 | NM_052964.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400168928.hg.1 | TC0400010050.hg.1 | Clariom D TC |

CLNK
Genomic Map