This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00402451_m1
See other SDR16C5 GE Assays ›
Gene Symbol:
SDR16C5
Gene Name:
short chain dehydrogenase/reductase family 16C, member 5
Gene Aliases:
EPHD-2, RDH#2, RDH-E2, RDHE2, retSDR2
Chromosome Location:
Chr.8: 56299648 - 56320776 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00402451_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SDR16C5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001318049.1 NP_001304978.1 2 - 3 1060 65
NM_138969.3 NP_620419.2 2 - 3 1060 65
GenBank mRNA AK057667.1 - 2 - 3 402 65
AK095159.1 - 2 - 3 1060 65
BC037219.1 - 2 - 3 407 5181472 65
BC064525.1 - 2 - 3 379 5752917 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0800010511.hg.1
This assay targets the same location as HTA transcript cluster(s) TC08001232.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SDR16C5

Gene Ontology Categories:

Function(s) Process(es)

retinol metabolic process
retinal metabolic process
keratinocyte proliferation
detection of light stimulus involved in visual perception
oxidation-reduction process
retinol dehydrogenase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR08015972.hg.1 TC08001232.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR08015972.hg.1 ENST00000303749.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08015972.hg.1 ENST00000522671.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08015972.hg.1 ENST00000538514.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08015972.hg.1 NM_138969.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 ENST00000303749.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 ENST00000303749.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 ENST00000303749.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 ENST00000303749.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 ENST00000303749.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 ENST00000303749.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 ENST00000303749.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 ENST00000303749.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 ENST00000303749.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 ENST00000303749.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 ENST00000303749.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 ENST00000303749.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 ENST00000396721.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 ENST00000396721.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 ENST00000396721.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 ENST00000396721.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 ENST00000396721.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 ENST00000396721.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 ENST00000396721.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 ENST00000396721.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 ENST00000396721.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 ENST00000396721.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 ENST00000396721.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 ENST00000522671.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 ENST00000522671.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 ENST00000522671.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 ENST00000522671.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 ENST00000522671.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 ENST00000522671.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 ENST00000522671.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 ENST00000522671.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 ENST00000522671.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 ENST00000522671.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 ENST00000522671.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 ENST00000522671.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 ENST00000522671.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 NM_138969.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 NM_138969.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 NM_138969.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 NM_138969.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 NM_138969.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 NM_138969.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 NM_138969.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 NM_138969.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 NM_138969.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 NM_138969.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 NM_138969.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 NM_138969.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010511.hg.1 NM_138969.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR08015971.hg.1 TC08001232.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR08015971.hg.1 ENST00000303749.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08015971.hg.1 ENST00000396721.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08015971.hg.1 ENST00000522671.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08015971.hg.1 ENST00000538514.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08015971.hg.1 NM_138969.3 Refseq
Affymetrix Human Clariom D Assay PSR0800171426.hg.1 ENST00000303749.7 Ensembl
Affymetrix Human Clariom D Assay PSR0800171426.hg.1 ENST00000522671.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800171426.hg.1 NM_138969.3 Refseq
Affymetrix Human Clariom D Assay PSR0800171426.hg.1 ENST00000303749.7 Ensembl
Affymetrix Human Clariom D Assay PSR0800171426.hg.1 ENST00000522671.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800171426.hg.1 NM_138969.3 Refseq
Affymetrix Human Clariom D Assay PSR0800171426.hg.1 TC0800010511.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0800010511.hg.1 ENST00000303749.7 Ensembl
Affymetrix Human Clariom D Assay TC0800010511.hg.1 ENST00000303749.7 Ensembl
Affymetrix Human Clariom D Assay TC0800010511.hg.1 ENST00000303749.7 Ensembl
Affymetrix Human Clariom D Assay TC0800010511.hg.1 ENST00000303749.7 Ensembl
Affymetrix Human Clariom D Assay TC0800010511.hg.1 ENST00000303749.7 Ensembl
Affymetrix Human Clariom D Assay TC0800010511.hg.1 ENST00000303749.7 Ensembl
Affymetrix Human Clariom D Assay TC0800010511.hg.1 ENST00000303749.7 Ensembl
Affymetrix Human Clariom D Assay TC0800010511.hg.1 ENST00000303749.7 Ensembl
Affymetrix Human Clariom D Assay TC0800010511.hg.1 ENST00000303749.7 Ensembl
Affymetrix Human Clariom D Assay TC0800010511.hg.1 ENST00000303749.7 Ensembl
Affymetrix Human Clariom D Assay TC0800010511.hg.1 ENST00000303749.7 Ensembl
Affymetrix Human Clariom D Assay TC0800010511.hg.1 ENST00000303749.7 Ensembl
Affymetrix Human Clariom D Assay TC0800010511.hg.1 ENST00000396721.6 Ensembl
Affymetrix Human Clariom D Assay TC0800010511.hg.1 ENST00000396721.6 Ensembl
Affymetrix Human Clariom D Assay TC0800010511.hg.1 ENST00000396721.6 Ensembl
Affymetrix Human Clariom D Assay TC0800010511.hg.1 ENST00000396721.6 Ensembl
Affymetrix Human Clariom D Assay TC0800010511.hg.1 ENST00000396721.6 Ensembl
Affymetrix Human Clariom D Assay TC0800010511.hg.1 ENST00000396721.6 Ensembl
Affymetrix Human Clariom D Assay TC0800010511.hg.1 ENST00000396721.6 Ensembl
Affymetrix Human Clariom D Assay TC0800010511.hg.1 ENST00000396721.6 Ensembl
Affymetrix Human Clariom D Assay TC0800010511.hg.1 ENST00000396721.6 Ensembl
Affymetrix Human Clariom D Assay TC0800010511.hg.1 ENST00000396721.6 Ensembl
Affymetrix Human Clariom D Assay TC0800010511.hg.1 ENST00000396721.6 Ensembl
Affymetrix Human Clariom D Assay TC0800010511.hg.1 ENST00000522671.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010511.hg.1 ENST00000522671.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010511.hg.1 ENST00000522671.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010511.hg.1 ENST00000522671.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010511.hg.1 ENST00000522671.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010511.hg.1 ENST00000522671.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010511.hg.1 ENST00000522671.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010511.hg.1 ENST00000522671.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010511.hg.1 ENST00000522671.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010511.hg.1 ENST00000522671.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010511.hg.1 ENST00000522671.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010511.hg.1 ENST00000522671.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010511.hg.1 ENST00000522671.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010511.hg.1 NM_138969.3 Refseq
Affymetrix Human Clariom D Assay TC0800010511.hg.1 NM_138969.3 Refseq
Affymetrix Human Clariom D Assay TC0800010511.hg.1 NM_138969.3 Refseq
Affymetrix Human Clariom D Assay TC0800010511.hg.1 NM_138969.3 Refseq
Affymetrix Human Clariom D Assay TC0800010511.hg.1 NM_138969.3 Refseq
Affymetrix Human Clariom D Assay TC0800010511.hg.1 NM_138969.3 Refseq
Affymetrix Human Clariom D Assay TC0800010511.hg.1 NM_138969.3 Refseq
Affymetrix Human Clariom D Assay TC0800010511.hg.1 NM_138969.3 Refseq
Affymetrix Human Clariom D Assay TC0800010511.hg.1 NM_138969.3 Refseq
Affymetrix Human Clariom D Assay TC0800010511.hg.1 NM_138969.3 Refseq
Affymetrix Human Clariom D Assay TC0800010511.hg.1 NM_138969.3 Refseq
Affymetrix Human Clariom D Assay TC0800010511.hg.1 NM_138969.3 Refseq
Affymetrix Human Clariom D Assay TC0800010511.hg.1 NM_138969.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0