This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00404261_m1
See other PTPN20 GE Assays ›
Gene Symbol:
PTPN20
Gene Name:
protein tyrosine phosphatase, non-receptor type 20
Gene Aliases:
CT126, PTPN20A, PTPN20B, bA142I17.1, bA42B19.1
Chromosome Location:
Chr.10: 46911134 - 47003834 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00404261_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PTPN20
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001042357.4 NP_001035816.1 10 - 11 1426 89
NM_001042358.4 NP_001035817.1 9 - 10 1284 89
NM_001042359.3 NP_001035818.1 7 - 8 858 89
NM_001042360.3 NP_001035819.1 4 - 5 735 89
NM_001042361.4 NP_001035820.1 7 - 8 733 89
NM_001042362.3 NP_001035821.1 5 - 6 490 89
NM_001042363.4 NP_001035822.1 9 - 10 1269 89
NM_001042364.4 NP_001035823.1 6 - 7 623 89
NM_001042365.3 NP_001035824.1 3 - 4 282 89
NM_001320681.1 NP_001307610.1 8 - 9 1174 89
NM_001320682.1 NP_001307611.1 4 - 5 380 89
NM_001320683.1 NP_001307612.1 8 - 9 890 89
NM_001320684.1 NP_001307613.1 7 - 8 799 89
NM_001320685.1 NP_001307614.1 9 - 10 1346 89
NM_001320686.1 NP_001307615.1 8 - 9 1189 89
NM_001320688.1 NP_001307617.1 6 - 7 763 89
NM_001320689.1 NP_001307618.1 5 - 6 531 89
NM_001320690.1 NP_001307619.1 4 - 5 395 89
NM_001320691.1 NP_001307620.1 7 - 8 1098 89
NM_015605.8 NP_056420.3 8 - 9 875 89
XM_011539605.2 XP_011537907.1 10 - 11 1570 89
XM_011539606.2 XP_011537908.1 10 - 11 2662 89
XM_011539607.2 XP_011537909.1 10 - 11 1561 89
XM_011539610.2 XP_011537912.1 7 - 8 1044 89
XM_011539611.2 XP_011537913.1 6 - 7 1044 89
XM_017016044.1 XP_016871533.1 9 - 10 1327 89
XM_017016045.1 XP_016871534.1 4 - 5 2646 89
XM_017016046.1 XP_016871535.1 9 - 10 1189 89
XM_017016049.1 XP_016871538.1 7 - 8 920 89
XM_017016050.1 XP_016871539.1 5 - 6 787 89
XM_017016053.1 XP_016871542.1 7 - 8 763 89
XM_017016054.1 XP_016871543.1 7 - 8 776 89
XM_017016056.1 XP_016871545.1 6 - 7 771 89
XM_017016057.1 XP_016871546.1 7 - 8 1081 89
XM_017016058.1 XP_016871547.1 5 - 6 807 89
XM_017016059.1 XP_016871548.1 6 - 7 765 89
XM_017016060.1 XP_016871549.1 6 - 7 611 89
XM_017016061.1 XP_016871550.1 5 - 6 561 89
XM_017016062.1 XP_016871551.1 5 - 6 681 89
XM_017016063.1 XP_016871552.1 6 - 7 652 89
XM_017016064.1 XP_016871553.1 4 - 5 404 89
XM_017016065.1 XP_016871554.1 5 - 6 454 89
GenBank mRNA AL050040.1 - 8 - 9 1132 89
AY704141.1 - 9 - 10 1225 89
AY704142.1 - 7 - 8 799 89
AY704143.1 - 4 - 5 676 89
AY704144.1 - 7 - 8 674 89
AY704145.1 - 5 - 6 431 89
AY704146.1 - 9 - 10 1274 89
AY704147.1 - 8 - 9 831 89
AY704148.1 - 7 - 8 740 89
AY704149.1 - 5 - 6 480 89
AY704150.1 - 9 - 10 1287 89
AY704151.1 - 6 - 7 628 89
AY704152.1 - 8 - 9 1130 89
AY704153.1 - 6 - 7 704 89
AY704154.1 - 6 - 7 579 89
AY704155.1 - 5 - 6 521 89
AY704156.1 - 5 - 6 472 89
AY704157.1 - 4 - 5 336 89
AY704158.1 - 7 - 8 1039 89
BC093750.1 - 7 - 8 729 7939595 89
BC113473.1 - 7 - 8 729 8322525 89
BC143394.1 - 5 - 6 486 89
BC171725.1 - 5 - 6 486 89
BX648913.1 - 3 - 4 283 89
CR933668.1 - 4 - 5 348 89

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PTPN20

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

peptidyl-tyrosine dephosphorylation
protein tyrosine phosphatase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline