This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00404273_m1
See other SMIM8 GE Assays ›
Gene Symbol:
SMIM8
Gene Name:
small integral membrane protein 8
Gene Aliases:
C6orf162, dJ102H19.2
Chromosome Location:
Chr.6: 87322588 - 87342328 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00404273_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SMIM8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001042493.2 NP_001035958.1 3 - 4 223 77
NM_020425.5 NP_065158.3 2 - 3 202 77
GenBank mRNA AF311339.1 - 2 - 3 244 77
AK311963.1 - 3 - 4 194 77
AL050201.1 - 1 - 2 169 77
BC070260.1 - 2 - 3 184 6452586 77
BG534658.1 - 2 - 3 203 77
DA293899.1 - 3 - 4 194 77

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SMIM8

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline