This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00410670_m1
See other PTPRK GE Assays ›
Gene Symbol:
PTPRK
Gene Name:
protein tyrosine phosphatase, receptor type K
Gene Aliases:
R-PTP-kappa
Chromosome Location:
Chr.6: 127968779 - 128520847 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00410670_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PTPRK
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001135648.2 NP_001129120.1 15 - 16 2810 71
NM_001291984.1 NP_001278913.1 15 - 16 2807 71
NM_002844.3 NP_002835.2 15 - 16 2810 71
XM_006715537.2 XP_006715600.1 15 - 16 2858 71
XM_017011145.1 XP_016866634.1 16 - 17 2825 71
XM_017011146.1 XP_016866635.1 16 - 17 2774 71
XM_017011147.1 XP_016866636.1 16 - 17 2752 71
XM_017011148.1 XP_016866637.1 16 - 17 2749 71
GenBank mRNA AF533875.1 - 15 - 16 2575 71
BC140775.1 - 15 - 16 2612 9021643 71
BC144512.1 - 15 - 16 2612 9053039 71
L77886.1 - 15 - 16 2509 71
Z70660.1 - 15 - 16 2491 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600013126.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06002092.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PTPRK

Panther Classification:

Molecular Function -

extracellular matrix glycoprotein

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
cell adhesion
signal transduction
transforming growth factor beta receptor signaling pathway
negative regulation of cell proliferation
negative regulation of keratinocyte proliferation
cell migration
negative regulation of cell migration
neuron projection development
protein localization to cell surface
cellular response to reactive oxygen species
cellular response to UV
peptidyl-tyrosine dephosphorylation
negative regulation of cell cycle
negative regulation of transcription, DNA-templated
focal adhesion assembly
protein tyrosine phosphatase activity
transmembrane receptor protein tyrosine phosphatase activity
protein binding
beta-catenin binding
protein kinase binding
gamma-catenin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR06026206.hg.1 TC06002092.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR06026206.hg.1 ENST00000368207.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06026206.hg.1 ENST00000368210.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06026206.hg.1 ENST00000368213.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06026206.hg.1 ENST00000368215.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06026206.hg.1 ENST00000368226.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06026206.hg.1 ENST00000368227.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06026206.hg.1 ENST00000532331.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06026206.hg.1 NM_001135648.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06026206.hg.1 NM_002844.3 Refseq
Affymetrix Human Clariom D Assay PSR0600190541.hg.1 ENST00000368207.7 Ensembl
Affymetrix Human Clariom D Assay PSR0600190541.hg.1 ENST00000368210.7 Ensembl
Affymetrix Human Clariom D Assay PSR0600190541.hg.1 ENST00000368213.9 Ensembl
Affymetrix Human Clariom D Assay PSR0600190541.hg.1 ENST00000368215.7 Ensembl
Affymetrix Human Clariom D Assay PSR0600190541.hg.1 ENST00000368226.8 Ensembl
Affymetrix Human Clariom D Assay PSR0600190541.hg.1 ENST00000415046.6 Ensembl
Affymetrix Human Clariom D Assay PSR0600190541.hg.1 ENST00000415055.2 Ensembl
Affymetrix Human Clariom D Assay PSR0600190541.hg.1 ENST00000532331.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600190541.hg.1 NM_001291984.1 Refseq
Affymetrix Human Clariom D Assay PSR0600190541.hg.1 NM_002844.3 Refseq
Affymetrix Human Clariom D Assay PSR0600190541.hg.1 ENST00000368207.7 Ensembl
Affymetrix Human Clariom D Assay PSR0600190541.hg.1 ENST00000368210.7 Ensembl
Affymetrix Human Clariom D Assay PSR0600190541.hg.1 ENST00000368213.9 Ensembl
Affymetrix Human Clariom D Assay PSR0600190541.hg.1 ENST00000368215.7 Ensembl
Affymetrix Human Clariom D Assay PSR0600190541.hg.1 ENST00000368226.8 Ensembl
Affymetrix Human Clariom D Assay PSR0600190541.hg.1 ENST00000415046.6 Ensembl
Affymetrix Human Clariom D Assay PSR0600190541.hg.1 ENST00000415055.2 Ensembl
Affymetrix Human Clariom D Assay PSR0600190541.hg.1 ENST00000532331.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600190541.hg.1 NM_001291984.1 Refseq
Affymetrix Human Clariom D Assay PSR0600190541.hg.1 NM_002844.3 Refseq
Affymetrix Human Clariom D Assay PSR0600190541.hg.1 TC0600013126.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0