This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00414418_m1
See other ECE2 GE Assays ›
Gene Symbol:
ECE2
Gene Name:
endothelin converting enzyme 2
Gene Aliases:
-
Chromosome Location:
Chr.3: 184249657 - 184293031 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00414418_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ECE2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_014693.3 NP_055508.3 1 - 2 231 53
NM_032331.3 NP_115707.2 1 - 2 231 53
GenBank mRNA AF428263.1 - 1 - 2 159 53
BC005835.2 - 1 - 2 227 53
BC012449.1 - 1 - 2 204 53
BC069005.1 - 1 - 2 217 53
BU568980.1 - 234 53

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF428263.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ECE2

Panther Classification:

Molecular Function -

metalloprotease methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

cell-cell signaling
brain development
heart development
cardioblast differentiation
peptide hormone processing
methylation
metalloendopeptidase activity
protein binding
methyltransferase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0