This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00415531_m1
See other VSIG1 GE Assays ›
Gene Symbol:
VSIG1
Gene Name:
V-set and immunoglobulin domain containing 1
Gene Aliases:
1700062D20Rik, GPA34, dJ889N15.1
Chromosome Location:
Chr.X: 108018886 - 108079184 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00415531_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: VSIG1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_182607.4 NP_872413.1 2 - 3 366 76
GenBank mRNA AK292472.1 - 2 - 3 245 76
AK301783.1 - 2 - 3 365 76
AK315846.1 - 2 - 3 271 76
BC043216.2 - 2 - 3 322 5295297 76
BX640913.1 - 2 - 3 280 76
BX648658.1 - 2 - 3 366 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0X00008080.hg.1
This assay targets the same location as HTA transcript cluster(s) TC0X000529.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene VSIG1

Panther Classification:

Molecular Function -

defense/immunity protein immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s)

epithelial cell morphogenesis
maintenance of gastrointestinal epithelium

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR0X007072.hg.1 TC0X000529.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR0X007072.hg.1 ENST00000217957.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X007072.hg.1 ENST00000415430.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X007072.hg.1 NM_001170553.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X007072.hg.1 NM_182607.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X007077.hg.1 TC0X000529.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR0X007077.hg.1 ENST00000217957.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X007077.hg.1 ENST00000415430.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X007077.hg.1 ENST00000485533.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X007077.hg.1 NM_001170553.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X007077.hg.1 NM_182607.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X007071.hg.1 TC0X000529.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR0X007071.hg.1 ENST00000415430.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X007071.hg.1 NM_001170553.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X007071.hg.1 NM_182607.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X007074.hg.1 TC0X000529.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR0X007074.hg.1 ENST00000217957.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X007074.hg.1 ENST00000415430.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X007074.hg.1 NM_001170553.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X007074.hg.1 NM_182607.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X007073.hg.1 TC0X000529.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR0X007073.hg.1 ENST00000217957.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X007073.hg.1 ENST00000415430.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X007073.hg.1 NM_001170553.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X007073.hg.1 NM_182607.4 Refseq
Affymetrix Human Clariom D Assay PSR0X00155243.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Clariom D Assay PSR0X00155243.hg.1 NM_001170553.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00155243.hg.1 NM_182607.4 Refseq
Affymetrix Human Clariom D Assay PSR0X00155243.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Clariom D Assay PSR0X00155243.hg.1 NM_001170553.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00155243.hg.1 NM_182607.4 Refseq
Affymetrix Human Clariom D Assay PSR0X00155243.hg.1 TC0X00008080.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0X00155250.hg.1 ENST00000217957.9 Ensembl
Affymetrix Human Clariom D Assay PSR0X00155250.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Clariom D Assay PSR0X00155250.hg.1 ENST00000458383.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00155250.hg.1 ENST00000485533.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00155250.hg.1 NM_001170553.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00155250.hg.1 NM_182607.4 Refseq
Affymetrix Human Clariom D Assay PSR0X00155250.hg.1 ENST00000217957.9 Ensembl
Affymetrix Human Clariom D Assay PSR0X00155250.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Clariom D Assay PSR0X00155250.hg.1 ENST00000458383.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00155250.hg.1 ENST00000485533.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00155250.hg.1 NM_001170553.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00155250.hg.1 NM_182607.4 Refseq
Affymetrix Human Clariom D Assay PSR0X00155250.hg.1 TC0X00008080.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0X00155247.hg.1 ENST00000217957.9 Ensembl
Affymetrix Human Clariom D Assay PSR0X00155247.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Clariom D Assay PSR0X00155247.hg.1 ENST00000458383.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00155247.hg.1 NM_001170553.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00155247.hg.1 NM_182607.4 Refseq
Affymetrix Human Clariom D Assay PSR0X00155247.hg.1 ENST00000217957.9 Ensembl
Affymetrix Human Clariom D Assay PSR0X00155247.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Clariom D Assay PSR0X00155247.hg.1 ENST00000458383.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00155247.hg.1 NM_001170553.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00155247.hg.1 NM_182607.4 Refseq
Affymetrix Human Clariom D Assay PSR0X00155247.hg.1 TC0X00008080.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0X00155246.hg.1 ENST00000217957.9 Ensembl
Affymetrix Human Clariom D Assay PSR0X00155246.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Clariom D Assay PSR0X00155246.hg.1 ENST00000458383.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00155246.hg.1 NM_001170553.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00155246.hg.1 NM_182607.4 Refseq
Affymetrix Human Clariom D Assay PSR0X00155246.hg.1 ENST00000217957.9 Ensembl
Affymetrix Human Clariom D Assay PSR0X00155246.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Clariom D Assay PSR0X00155246.hg.1 ENST00000458383.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00155246.hg.1 NM_001170553.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00155246.hg.1 NM_182607.4 Refseq
Affymetrix Human Clariom D Assay PSR0X00155246.hg.1 TC0X00008080.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0X00155244.hg.1 ENST00000217957.9 Ensembl
Affymetrix Human Clariom D Assay PSR0X00155244.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Clariom D Assay PSR0X00155244.hg.1 NM_001170553.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00155244.hg.1 NM_182607.4 Refseq
Affymetrix Human Clariom D Assay PSR0X00155244.hg.1 ENST00000217957.9 Ensembl
Affymetrix Human Clariom D Assay PSR0X00155244.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Clariom D Assay PSR0X00155244.hg.1 NM_001170553.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00155244.hg.1 NM_182607.4 Refseq
Affymetrix Human Clariom D Assay PSR0X00155244.hg.1 TC0X00008080.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0X00155245.hg.1 ENST00000217957.9 Ensembl
Affymetrix Human Clariom D Assay PSR0X00155245.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Clariom D Assay PSR0X00155245.hg.1 ENST00000458383.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00155245.hg.1 NM_001170553.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00155245.hg.1 NM_182607.4 Refseq
Affymetrix Human Clariom D Assay PSR0X00155245.hg.1 ENST00000217957.9 Ensembl
Affymetrix Human Clariom D Assay PSR0X00155245.hg.1 ENST00000415430.7 Ensembl
Affymetrix Human Clariom D Assay PSR0X00155245.hg.1 ENST00000458383.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00155245.hg.1 NM_001170553.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00155245.hg.1 NM_182607.4 Refseq
Affymetrix Human Clariom D Assay PSR0X00155245.hg.1 TC0X00008080.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 147224 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0