This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00416100_m1
See other LINC00400 GE Assays ›
Gene Symbol:
LINC00400
Gene Name:
long intergenic non-protein coding RNA 400
Gene Aliases:
-
Chromosome Location:
Chr.13: 43113165 - 43159466 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00416100_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LINC00400
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_047013.1 1 - 2 117 108
GenBank mRNA AV711458.1 - 1 - 2 117 108

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Homo sapiens Inventoried Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-7wnc4:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline