This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00416967_g1
See other H1FOO GE Assays ›
Gene Symbol:
H1FOO
Gene Name:
H1 histone family member O, oocyte specific
Gene Aliases:
H1.8, H1oo, osH1
Chromosome Location:
Chr.3: 129543214 - 129555185 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00416967_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: H1FOO
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001308262.1 NP_001295191.1 1 - 2 95 62
GenBank mRNA BC047943.1 - 1 - 2 94 5742122 62
BM564157.1 - 112 62

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC047943.1, NM_001308262.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

Panther Classification:

Molecular Function -

histone

Gene Ontology Categories:

Function(s) Process(es)

nucleosome assembly
nucleosome positioning
regulation of DNA methylation
meiotic cell cycle
negative regulation of stem cell differentiation
nucleosomal DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300008789.hg.1 ENST00000324382.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008789.hg.1 ENST00000324382.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008789.hg.1 ENST00000324382.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008789.hg.1 ENST00000324382.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008789.hg.1 ENST00000324382.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008789.hg.1 ENST00000503977.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008789.hg.1 ENST00000503977.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008789.hg.1 ENST00000503977.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008789.hg.1 ENST00000503977.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008789.hg.1 ENST00000503977.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008789.hg.1 NM_001308262.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008789.hg.1 NM_001308262.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008789.hg.1 NM_001308262.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008789.hg.1 NM_001308262.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008789.hg.1 NM_001308262.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008789.hg.1 NM_153833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008789.hg.1 NM_153833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008789.hg.1 NM_153833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008789.hg.1 NM_153833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008789.hg.1 NM_153833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008789.hg.1 NM_153833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400009754.hg.1 ENST00000557720.2 Ensembl
Affymetrix Human Clariom D Assay TC1400009754.hg.1 ENST00000557720.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300162529.hg.1 ENST00000503977.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300162529.hg.1 NM_001308262.1 Refseq
Affymetrix Human Clariom D Assay PSR0300162529.hg.1 ENST00000503977.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300162529.hg.1 NM_001308262.1 Refseq
Affymetrix Human Clariom D Assay PSR0300162529.hg.1 TC0300008789.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0300008789.hg.1 ENST00000324382.6 Ensembl
Affymetrix Human Clariom D Assay TC0300008789.hg.1 ENST00000324382.6 Ensembl
Affymetrix Human Clariom D Assay TC0300008789.hg.1 ENST00000324382.6 Ensembl
Affymetrix Human Clariom D Assay TC0300008789.hg.1 ENST00000324382.6 Ensembl
Affymetrix Human Clariom D Assay TC0300008789.hg.1 ENST00000324382.6 Ensembl
Affymetrix Human Clariom D Assay TC0300008789.hg.1 ENST00000503977.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008789.hg.1 ENST00000503977.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008789.hg.1 ENST00000503977.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008789.hg.1 ENST00000503977.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008789.hg.1 ENST00000503977.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008789.hg.1 NM_001308262.1 Refseq
Affymetrix Human Clariom D Assay TC0300008789.hg.1 NM_001308262.1 Refseq
Affymetrix Human Clariom D Assay TC0300008789.hg.1 NM_001308262.1 Refseq
Affymetrix Human Clariom D Assay TC0300008789.hg.1 NM_001308262.1 Refseq
Affymetrix Human Clariom D Assay TC0300008789.hg.1 NM_001308262.1 Refseq
Affymetrix Human Clariom D Assay TC0300008789.hg.1 NM_153833.2 Refseq
Affymetrix Human Clariom D Assay TC0300008789.hg.1 NM_153833.2 Refseq
Affymetrix Human Clariom D Assay TC0300008789.hg.1 NM_153833.2 Refseq
Affymetrix Human Clariom D Assay TC0300008789.hg.1 NM_153833.2 Refseq
Affymetrix Human Clariom D Assay TC0300008789.hg.1 NM_153833.2 Refseq
Affymetrix Human Clariom D Assay TC0300008789.hg.1 NM_153833.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline