This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00416984_m1
See other HACD4 GE Assays ›
Gene Symbol:
HACD4
Gene Name:
3-hydroxyacyl-CoA dehydratase 4
Gene Aliases:
PTPLAD2
Chromosome Location:
Chr.9: 21003621 - 21031712 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00416984_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HACD4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001010915.4 NP_001010915.2 1 - 2 121 88
NM_001321903.1 NP_001308832.1 1 - 2 121 88
XM_017014714.1 XP_016870203.1 1 - 2 116 88
GenBank mRNA BC114215.1 - 1 - 2 55 5174279 88
BX648779.1 - 1 - 2 83 88
HY118500.1 - 1 - 2 121 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900009685.hg.1
This assay targets the same location as HTA transcript cluster(s) TC09000945.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene HACD4

Gene Ontology Categories:

Function(s) Process(es)

fatty acid elongation
very long-chain fatty acid biosynthetic process
3-hydroxyacyl-CoA dehydratase activity
enzyme binding
3-hydroxy-behenoyl-CoA dehydratase activity
3-hydroxy-lignoceroyl-CoA dehydratase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR09012650.hg.1 TC09000945.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR09012650.hg.1 ENST00000495827.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09012650.hg.1 ENST00000513293.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09012650.hg.1 NM_001010915.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 ENST00000488436.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 ENST00000488436.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 ENST00000488436.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 ENST00000488436.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 NM_001010915.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 NM_001010915.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 NM_001010915.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 NM_001010915.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 NM_001010915.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 NM_001010915.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 NM_001010915.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 NM_001010915.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900009685.hg.1 NM_001010915.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09012651.hg.1 TC09000945.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR09012651.hg.1 ENST00000495827.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09012651.hg.1 ENST00000513293.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09012651.hg.1 NM_001010915.4 Refseq
Affymetrix Human Clariom D Assay PSR0900166273.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900166273.hg.1 NM_001010915.4 Refseq
Affymetrix Human Clariom D Assay PSR0900166273.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900166273.hg.1 NM_001010915.4 Refseq
Affymetrix Human Clariom D Assay PSR0900166273.hg.1 TC0900009685.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0900166274.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900166274.hg.1 NM_001010915.4 Refseq
Affymetrix Human Clariom D Assay PSR0900166274.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900166274.hg.1 NM_001010915.4 Refseq
Affymetrix Human Clariom D Assay PSR0900166274.hg.1 TC0900009685.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0900166276.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900166276.hg.1 NM_001010915.4 Refseq
Affymetrix Human Clariom D Assay PSR0900166276.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Clariom D Assay PSR0900166276.hg.1 NM_001010915.4 Refseq
Affymetrix Human Clariom D Assay PSR0900166276.hg.1 TC0900009685.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0900009685.hg.1 ENST00000488436.1 Ensembl
Affymetrix Human Clariom D Assay TC0900009685.hg.1 ENST00000488436.1 Ensembl
Affymetrix Human Clariom D Assay TC0900009685.hg.1 ENST00000488436.1 Ensembl
Affymetrix Human Clariom D Assay TC0900009685.hg.1 ENST00000488436.1 Ensembl
Affymetrix Human Clariom D Assay TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009685.hg.1 ENST00000495827.2 Ensembl
Affymetrix Human Clariom D Assay TC0900009685.hg.1 NM_001010915.4 Refseq
Affymetrix Human Clariom D Assay TC0900009685.hg.1 NM_001010915.4 Refseq
Affymetrix Human Clariom D Assay TC0900009685.hg.1 NM_001010915.4 Refseq
Affymetrix Human Clariom D Assay TC0900009685.hg.1 NM_001010915.4 Refseq
Affymetrix Human Clariom D Assay TC0900009685.hg.1 NM_001010915.4 Refseq
Affymetrix Human Clariom D Assay TC0900009685.hg.1 NM_001010915.4 Refseq
Affymetrix Human Clariom D Assay TC0900009685.hg.1 NM_001010915.4 Refseq
Affymetrix Human Clariom D Assay TC0900009685.hg.1 NM_001010915.4 Refseq
Affymetrix Human Clariom D Assay TC0900009685.hg.1 NM_001010915.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0