This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00417718_m1
See other CPTP GE Assays ›
Gene Symbol:
CPTP
Gene Name:
ceramide-1-phosphate transfer protein
Gene Aliases:
GLTPD1
Chromosome Location:
Chr.1: 1324756 - 1328896 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00417718_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CPTP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005244801.3 XP_005244858.1 1 - 2 452 128

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_005244801.3 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Inventoried Probe spans exons

Panther Classification:

Molecular Function -

transfer/carrier protein

Gene Ontology Categories:

Function(s) Process(es)

glycosphingolipid metabolic process
glycolipid transport
ceramide 1-phosphate transport
phospholipid binding
phospholipid transporter activity
glycolipid binding
ceramide 1-phosphate binding
ceramide 1-phosphate transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000464957.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000464957.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000464957.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000464957.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000488011.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000488011.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000488011.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000488011.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 NM_001029885.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 NM_001029885.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 NM_001029885.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 NM_001029885.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 NM_001029885.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 NM_001029885.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 NM_001029885.1 Refseq
Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000464957.1 Ensembl
Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000464957.1 Ensembl
Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000464957.1 Ensembl
Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000464957.1 Ensembl
Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000488011.1 Ensembl
Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000488011.1 Ensembl
Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000488011.1 Ensembl
Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000488011.1 Ensembl
Affymetrix Human Clariom D Assay TC0100006501.hg.1 NM_001029885.1 Refseq
Affymetrix Human Clariom D Assay TC0100006501.hg.1 NM_001029885.1 Refseq
Affymetrix Human Clariom D Assay TC0100006501.hg.1 NM_001029885.1 Refseq
Affymetrix Human Clariom D Assay TC0100006501.hg.1 NM_001029885.1 Refseq
Affymetrix Human Clariom D Assay TC0100006501.hg.1 NM_001029885.1 Refseq
Affymetrix Human Clariom D Assay TC0100006501.hg.1 NM_001029885.1 Refseq
Affymetrix Human Clariom D Assay TC0100006501.hg.1 NM_001029885.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0