This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00417719_g1
See other CPTP GE Assays ›
Gene Symbol:
CPTP
Gene Name:
ceramide-1-phosphate transfer protein
Gene Aliases:
GLTPD1
Chromosome Location:
Chr.1: 1324756 - 1328896 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00417719_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CPTP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001029885.1 NP_001025056.1 2 - 3 538 56
XM_005244801.3 XP_005244858.1 2 - 3 651 56
XM_011542200.2 XP_011540502.1 2 - 3 547 56
GenBank mRNA JN542538.1 - 1 - 2 123 56

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100006501.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01000034.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CPTP

Panther Classification:

Molecular Function -

transfer/carrier protein

Gene Ontology Categories:

Function(s) Process(es)

glycosphingolipid metabolic process
glycolipid transport
ceramide 1-phosphate transport
phospholipid binding
phospholipid transporter activity
glycolipid binding
ceramide 1-phosphate binding
ceramide 1-phosphate transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01000427.hg.1 TC01000034.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01000427.hg.1 ENST00000343938.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01000427.hg.1 ENST00000464957.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01000427.hg.1 NM_001029885.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01000431.hg.1 TC01000034.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01000431.hg.1 ENST00000343938.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01000431.hg.1 ENST00000464957.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01000431.hg.1 NM_001029885.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01000429.hg.1 TC01000034.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01000429.hg.1 ENST00000343938.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01000429.hg.1 NM_001029885.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01000430.hg.1 TC01000034.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01000430.hg.1 ENST00000343938.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01000430.hg.1 ENST00000464957.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01000430.hg.1 NM_001029885.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000464957.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000464957.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000464957.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000464957.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000488011.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000488011.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000488011.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 ENST00000488011.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 NM_001029885.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 NM_001029885.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 NM_001029885.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 NM_001029885.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 NM_001029885.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 NM_001029885.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006501.hg.1 NM_001029885.1 Refseq
Affymetrix Human Clariom D Assay PSR0100144762.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100144762.hg.1 ENST00000464957.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100144762.hg.1 NM_001029885.1 Refseq
Affymetrix Human Clariom D Assay PSR0100144762.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100144762.hg.1 ENST00000464957.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100144762.hg.1 NM_001029885.1 Refseq
Affymetrix Human Clariom D Assay PSR0100144762.hg.1 TC0100006501.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0100144760.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100144760.hg.1 ENST00000488011.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100144760.hg.1 NM_001029885.1 Refseq
Affymetrix Human Clariom D Assay PSR0100144760.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100144760.hg.1 ENST00000488011.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100144760.hg.1 NM_001029885.1 Refseq
Affymetrix Human Clariom D Assay PSR0100144760.hg.1 TC0100006501.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0100144759.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100144759.hg.1 ENST00000488011.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100144759.hg.1 NM_001029885.1 Refseq
Affymetrix Human Clariom D Assay PSR0100144759.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100144759.hg.1 ENST00000488011.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100144759.hg.1 NM_001029885.1 Refseq
Affymetrix Human Clariom D Assay PSR0100144759.hg.1 TC0100006501.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0100144757.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100144757.hg.1 ENST00000464957.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100144757.hg.1 NM_001029885.1 Refseq
Affymetrix Human Clariom D Assay PSR0100144757.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100144757.hg.1 ENST00000464957.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100144757.hg.1 NM_001029885.1 Refseq
Affymetrix Human Clariom D Assay PSR0100144757.hg.1 TC0100006501.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000464957.1 Ensembl
Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000464957.1 Ensembl
Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000464957.1 Ensembl
Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000464957.1 Ensembl
Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000488011.1 Ensembl
Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000488011.1 Ensembl
Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000488011.1 Ensembl
Affymetrix Human Clariom D Assay TC0100006501.hg.1 ENST00000488011.1 Ensembl
Affymetrix Human Clariom D Assay TC0100006501.hg.1 NM_001029885.1 Refseq
Affymetrix Human Clariom D Assay TC0100006501.hg.1 NM_001029885.1 Refseq
Affymetrix Human Clariom D Assay TC0100006501.hg.1 NM_001029885.1 Refseq
Affymetrix Human Clariom D Assay TC0100006501.hg.1 NM_001029885.1 Refseq
Affymetrix Human Clariom D Assay TC0100006501.hg.1 NM_001029885.1 Refseq
Affymetrix Human Clariom D Assay TC0100006501.hg.1 NM_001029885.1 Refseq
Affymetrix Human Clariom D Assay TC0100006501.hg.1 NM_001029885.1 Refseq
Affymetrix Human Clariom D Assay PSR0100144761.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100144761.hg.1 ENST00000464957.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100144761.hg.1 ENST00000488011.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100144761.hg.1 NM_001029885.1 Refseq
Affymetrix Human Clariom D Assay PSR0100144761.hg.1 ENST00000343938.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100144761.hg.1 ENST00000464957.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100144761.hg.1 ENST00000488011.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100144761.hg.1 NM_001029885.1 Refseq
Affymetrix Human Clariom D Assay PSR0100144761.hg.1 TC0100006501.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0