This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00420102_g1
See other LIPT2 GE Assays ›
Gene Symbol:
LIPT2
Gene Name:
lipoyl(octanoyl) transferase 2 (putative)
Gene Aliases:
-
Chromosome Location:
Chr.11: 74490727 - 74493972 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00420102_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LIPT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001144869.1 NP_001138341.1 1 - 2 477 68
GenBank mRNA CX752266.1 - 1 - 2 459 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100011616.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11002083.hg.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens 1700 Inventoried Probe spans exons

Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

lipoate biosynthetic process
protein lipoylation
glyoxylate metabolic process
octanoyltransferase activity
ligase activity
lipoyl(octanoyl) transferase activity

MicroArray Probes:

Agilent Affymetrix ArrayStar ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Human Genome Microarray 4x44K v2 A_23_P24922 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_1945 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_1946 - -
Agilent Whole Human Genome Oligo Microarray 4x44K 36894 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 1513 - -
Agilent Human Genome CGH + SNP Microarray A_16_P02470028 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_23_P24922 - -
SurePrint G3 Human GE Microarray G4851A A_23_P24922 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28295357 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28295335 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16473866 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07877084 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28295326 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07877091 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07877079 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07877074 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07877095 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16473849 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07877101 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07877080 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16473863 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16473854 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28295374 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28295325 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07877075 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28295334 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07877102 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07877087 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07877081 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16473858 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07877097 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07877100 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28295344 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28295359 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28295351 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16473851 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07877086 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28295348 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07877099 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07877071 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28295337 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28295373 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28295354 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16473865 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28295352 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28295357 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28295374 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28295325 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07877102 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07877075 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28295352 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28295348 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16473866 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07877097 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07877081 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28295344 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07877086 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16473863 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07877099 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16473854 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07877074 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07877101 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28295326 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28295359 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07877100 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16473858 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16473851 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28295337 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28295334 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16473865 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07877087 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16473849 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07877071 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28295335 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07877091 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07877084 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07877080 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07877079 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28295373 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28295351 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28295354 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07877095 - -
Whole Human Genome Oligo Microarray G4112A A_23_P24922 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07877102 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07877073 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07877089 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07877071 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07877080 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07877082 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07877100 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16473872 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07877079 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07877070 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07877098 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07877090 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16473866 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07877095 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16473865 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07877072 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07877084 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07877093 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07877101 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07877086 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P16473870 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P07877075 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.0 ST Array 7950421 - -
Affymetrix Human Gene 1.0 ST Array 7950422 - -
Affymetrix Human Gene 1.1 ST Array 7950420 - -
Affymetrix Human Genome U219 Array 11737142_PM_at - -
Affymetrix Human Genome U219 Array 11737142_at - -
Affymetrix Human Transcriptome Array 2.0 PSR11024984.hg.1 ENST00000310109.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11024984.hg.1 NM_001144869.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11024984.hg.1 TC11002083.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR11024983.hg.1 ENST00000310109.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11024983.hg.1 NM_001144869.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11024983.hg.1 TC11002083.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1100011616.hg.1 ENST00000310109.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011616.hg.1 ENST00000310109.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011616.hg.1 ENST00000310109.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011616.hg.1 ENST00000310109.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011616.hg.1 ENST00000310109.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011616.hg.1 ENST00000310109.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011616.hg.1 ENST00000310109.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011616.hg.1 ENST00000310109.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011616.hg.1 ENST00000527115.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011616.hg.1 ENST00000527115.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011616.hg.1 ENST00000527115.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011616.hg.1 ENST00000527115.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011616.hg.1 ENST00000527115.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011616.hg.1 ENST00000527115.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011616.hg.1 ENST00000527115.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011616.hg.1 ENST00000528085.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011616.hg.1 ENST00000528085.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011616.hg.1 NM_001144869.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011616.hg.1 NM_001144869.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011616.hg.1 NM_001144869.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011616.hg.1 NM_001144869.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011616.hg.1 NM_001144869.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011616.hg.1 NM_001144869.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011616.hg.1 NM_001144869.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013070.hg.1 ENST00000526036.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013070.hg.1 ENST00000526036.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013070.hg.1 ENST00000526036.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013070.hg.1 ENST00000526036.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013070.hg.1 ENST00000526036.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013070.hg.1 ENST00000526036.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013070.hg.1 ENST00000612462.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100189707.hg.1 ENST00000310109.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100189707.hg.1 ENST00000527115.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100189707.hg.1 NM_001144869.2 Refseq
Affymetrix Human Clariom D Assay PSR1100189707.hg.1 ENST00000310109.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100189707.hg.1 ENST00000527115.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100189707.hg.1 NM_001144869.2 Refseq
Affymetrix Human Clariom D Assay PSR1100189707.hg.1 TC1100011616.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1100189710.hg.1 ENST00000310109.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100189710.hg.1 ENST00000527115.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100189710.hg.1 NM_001144869.2 Refseq
Affymetrix Human Clariom D Assay PSR1100189710.hg.1 ENST00000310109.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100189710.hg.1 ENST00000527115.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100189710.hg.1 NM_001144869.2 Refseq
Affymetrix Human Clariom D Assay PSR1100189710.hg.1 TC1100011616.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1100189711.hg.1 ENST00000310109.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100189711.hg.1 NM_001144869.2 Refseq
Affymetrix Human Clariom D Assay PSR1100189711.hg.1 ENST00000310109.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100189711.hg.1 NM_001144869.2 Refseq
Affymetrix Human Clariom D Assay PSR1100189711.hg.1 TC1100011616.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1100013070.hg.1 ENST00000526036.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013070.hg.1 ENST00000526036.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013070.hg.1 ENST00000526036.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013070.hg.1 ENST00000526036.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013070.hg.1 ENST00000526036.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013070.hg.1 ENST00000526036.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013070.hg.1 ENST00000612462.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100189708.hg.1 ENST00000310109.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100189708.hg.1 ENST00000527115.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100189708.hg.1 ENST00000528085.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100189708.hg.1 NM_001144869.2 Refseq
Affymetrix Human Clariom D Assay PSR1100189708.hg.1 ENST00000310109.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100189708.hg.1 ENST00000527115.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100189708.hg.1 ENST00000528085.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100189708.hg.1 NM_001144869.2 Refseq
Affymetrix Human Clariom D Assay PSR1100189708.hg.1 TC1100011616.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1100189712.hg.1 ENST00000310109.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100189712.hg.1 ENST00000528085.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100189712.hg.1 NM_001144869.2 Refseq
Affymetrix Human Clariom D Assay PSR1100189712.hg.1 ENST00000310109.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100189712.hg.1 ENST00000528085.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100189712.hg.1 NM_001144869.2 Refseq
Affymetrix Human Clariom D Assay PSR1100189712.hg.1 TC1100011616.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1100189706.hg.1 ENST00000310109.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100189706.hg.1 ENST00000527115.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100189706.hg.1 NM_001144869.2 Refseq
Affymetrix Human Clariom D Assay PSR1100189706.hg.1 ENST00000310109.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100189706.hg.1 ENST00000527115.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100189706.hg.1 NM_001144869.2 Refseq
Affymetrix Human Clariom D Assay PSR1100189706.hg.1 TC1100011616.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1100011616.hg.1 ENST00000310109.4 Ensembl
Affymetrix Human Clariom D Assay TC1100011616.hg.1 ENST00000310109.4 Ensembl
Affymetrix Human Clariom D Assay TC1100011616.hg.1 ENST00000310109.4 Ensembl
Affymetrix Human Clariom D Assay TC1100011616.hg.1 ENST00000310109.4 Ensembl
Affymetrix Human Clariom D Assay TC1100011616.hg.1 ENST00000310109.4 Ensembl
Affymetrix Human Clariom D Assay TC1100011616.hg.1 ENST00000310109.4 Ensembl
Affymetrix Human Clariom D Assay TC1100011616.hg.1 ENST00000310109.4 Ensembl
Affymetrix Human Clariom D Assay TC1100011616.hg.1 ENST00000310109.4 Ensembl
Affymetrix Human Clariom D Assay TC1100011616.hg.1 ENST00000527115.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011616.hg.1 ENST00000527115.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011616.hg.1 ENST00000527115.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011616.hg.1 ENST00000527115.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011616.hg.1 ENST00000527115.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011616.hg.1 ENST00000527115.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011616.hg.1 ENST00000527115.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011616.hg.1 ENST00000528085.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011616.hg.1 ENST00000528085.1 Ensembl
Affymetrix Human Clariom D Assay TC1100011616.hg.1 NM_001144869.2 Refseq
Affymetrix Human Clariom D Assay TC1100011616.hg.1 NM_001144869.2 Refseq
Affymetrix Human Clariom D Assay TC1100011616.hg.1 NM_001144869.2 Refseq
Affymetrix Human Clariom D Assay TC1100011616.hg.1 NM_001144869.2 Refseq
Affymetrix Human Clariom D Assay TC1100011616.hg.1 NM_001144869.2 Refseq
Affymetrix Human Clariom D Assay TC1100011616.hg.1 NM_001144869.2 Refseq
Affymetrix Human Clariom D Assay TC1100011616.hg.1 NM_001144869.2 Refseq
Affymetrix Human Clariom D Assay PSR1100189713.hg.1 ENST00000310109.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100189713.hg.1 NM_001144869.2 Refseq
Affymetrix Human Clariom D Assay PSR1100189713.hg.1 ENST00000310109.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100189713.hg.1 NM_001144869.2 Refseq
Affymetrix Human Clariom D Assay PSR1100189713.hg.1 TC1100011616.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
Arraystar Human LncRNA microarray V2.0 ASHG19A3A045494 - -
Arraystar Human LncRNA microarray V2.0 ASHG19A3A045494 - -
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 159317 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0