This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00426435_m1
See other HSD3B1 GE Assays ›
Gene Symbol:
HSD3B1
Gene Name:
hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 1
Gene Aliases:
3BETAHSD, HSD3B, HSDB3, HSDB3A, I, SDR11E1
Chromosome Location:
Chr.1: 119507112 - 119515060 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00426435_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HSD3B1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000862.2 NP_000853.1 2 - 3 289 72
XM_011541314.2 XP_011539616.1 2 - 3 330 72
XM_017001119.1 XP_016856608.1 2 - 3 253 72
GenBank mRNA AK291556.1 - 1 - 2 303 72
AL544564.3 - 2 - 3 289 72
AL550307.3 - 2 - 3 271 72
BC031999.1 - 1 - 2 231 4755300 72
M27137.1 - 1 - 2 147 72
M35493.1 - 1 - 2 174 72
S45679.1 - 2 - 3 275 72
X53321.1 - 1 - 2 174 72
X55997.1 - 1 - 2 144 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100009614.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01001040.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Human Androgen Panel 1700 Inventoried Human_Androgen Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene HSD3B1

Panther Classification:

Molecular Function -

dehydratase

Gene Ontology Categories:

Function(s) Process(es)

steroid biosynthetic process
androgen biosynthetic process
estrogen biosynthetic process
glucocorticoid biosynthetic process
mineralocorticoid biosynthetic process
oxidation-reduction process
3-beta-hydroxy-delta5-steroid dehydrogenase activity
steroid delta-isomerase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100009616.hg.1 ENST00000434856.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009616.hg.1 ENST00000434856.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009616.hg.1 ENST00000434856.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01016485.hg.1 TC01001040.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01016485.hg.1 ENST00000235547.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01016485.hg.1 ENST00000369413.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01016485.hg.1 NM_000862.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009614.hg.1 ENST00000369413.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009614.hg.1 ENST00000369413.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009614.hg.1 ENST00000369413.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009614.hg.1 ENST00000369413.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009614.hg.1 ENST00000369413.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009614.hg.1 ENST00000369413.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009614.hg.1 ENST00000369413.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009614.hg.1 ENST00000487520.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009614.hg.1 ENST00000487520.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009614.hg.1 ENST00000487520.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009614.hg.1 ENST00000492140.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009614.hg.1 ENST00000492140.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009614.hg.1 ENST00000492140.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009614.hg.1 ENST00000492140.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009614.hg.1 ENST00000492140.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009614.hg.1 ENST00000528909.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009614.hg.1 ENST00000528909.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009614.hg.1 ENST00000528909.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009614.hg.1 ENST00000528909.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009614.hg.1 ENST00000528909.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009614.hg.1 ENST00000528909.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009614.hg.1 ENST00000528909.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009614.hg.1 ENST00000531340.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009614.hg.1 ENST00000531340.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009614.hg.1 ENST00000531340.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009614.hg.1 NM_000862.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009614.hg.1 NM_000862.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009614.hg.1 NM_000862.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009614.hg.1 NM_000862.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009614.hg.1 NM_000862.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009614.hg.1 NM_000862.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009614.hg.1 NM_000862.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009615.hg.1 ENST00000514706.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009615.hg.1 ENST00000514706.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009615.hg.1 ENST00000514706.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009613.hg.1 ENST00000286193.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009613.hg.1 ENST00000286193.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009613.hg.1 ENST00000286193.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009612.hg.1 ENST00000457615.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009612.hg.1 ENST00000457615.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009612.hg.1 ENST00000457615.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01016489.hg.1 TC01001040.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01016489.hg.1 ENST00000235547.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01016489.hg.1 ENST00000369413.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01016489.hg.1 ENST00000528909.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01016489.hg.1 NM_000862.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01016491.hg.1 TC01001040.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01016491.hg.1 ENST00000235547.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01016491.hg.1 ENST00000369413.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01016491.hg.1 ENST00000528909.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01016491.hg.1 NM_000862.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01016493.hg.1 TC01001040.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01016493.hg.1 ENST00000235547.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01016493.hg.1 ENST00000369413.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01016493.hg.1 ENST00000528909.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01016493.hg.1 NM_000862.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009617.hg.1 ENST00000511509.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009617.hg.1 ENST00000511509.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009617.hg.1 ENST00000511509.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009617.hg.1 NR_033781.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009617.hg.1 NR_033781.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009617.hg.1 NR_033781.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009617.hg.1 NR_033781.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009617.hg.1 NR_033781.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009617.hg.1 NR_033781.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009617.hg.1 NR_033781.1 Refseq
Affymetrix Human Clariom D Assay PSR0100169201.hg.1 ENST00000369413.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100169201.hg.1 ENST00000492140.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100169201.hg.1 ENST00000528909.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100169201.hg.1 NM_000862.2 Refseq
Affymetrix Human Clariom D Assay PSR0100169201.hg.1 ENST00000369413.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100169201.hg.1 ENST00000492140.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100169201.hg.1 ENST00000528909.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100169201.hg.1 NM_000862.2 Refseq
Affymetrix Human Clariom D Assay PSR0100169201.hg.1 TC0100009614.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100009617.hg.1 ENST00000511509.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009617.hg.1 ENST00000511509.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009617.hg.1 ENST00000511509.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009617.hg.1 NR_033781.1 Refseq
Affymetrix Human Clariom D Assay TC0100009617.hg.1 NR_033781.1 Refseq
Affymetrix Human Clariom D Assay TC0100009617.hg.1 NR_033781.1 Refseq
Affymetrix Human Clariom D Assay TC0100009617.hg.1 NR_033781.1 Refseq
Affymetrix Human Clariom D Assay TC0100009617.hg.1 NR_033781.1 Refseq
Affymetrix Human Clariom D Assay TC0100009617.hg.1 NR_033781.1 Refseq
Affymetrix Human Clariom D Assay TC0100009617.hg.1 NR_033781.1 Refseq
Affymetrix Human Clariom D Assay TC0100009613.hg.1 ENST00000286193.2 Ensembl
Affymetrix Human Clariom D Assay TC0100009613.hg.1 ENST00000286193.2 Ensembl
Affymetrix Human Clariom D Assay TC0100009613.hg.1 ENST00000286193.2 Ensembl
Affymetrix Human Clariom D Assay PSR0100169200.hg.1 ENST00000369413.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100169200.hg.1 ENST00000492140.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100169200.hg.1 ENST00000528909.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100169200.hg.1 ENST00000531340.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100169200.hg.1 NM_000862.2 Refseq
Affymetrix Human Clariom D Assay PSR0100169200.hg.1 ENST00000369413.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100169200.hg.1 ENST00000492140.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100169200.hg.1 ENST00000528909.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100169200.hg.1 ENST00000531340.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100169200.hg.1 NM_000862.2 Refseq
Affymetrix Human Clariom D Assay PSR0100169200.hg.1 TC0100009614.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100009614.hg.1 ENST00000369413.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009614.hg.1 ENST00000369413.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009614.hg.1 ENST00000369413.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009614.hg.1 ENST00000369413.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009614.hg.1 ENST00000369413.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009614.hg.1 ENST00000369413.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009614.hg.1 ENST00000369413.7 Ensembl
Affymetrix Human Clariom D Assay TC0100009614.hg.1 ENST00000487520.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009614.hg.1 ENST00000487520.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009614.hg.1 ENST00000487520.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009614.hg.1 ENST00000492140.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009614.hg.1 ENST00000492140.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009614.hg.1 ENST00000492140.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009614.hg.1 ENST00000492140.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009614.hg.1 ENST00000492140.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009614.hg.1 ENST00000528909.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009614.hg.1 ENST00000528909.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009614.hg.1 ENST00000528909.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009614.hg.1 ENST00000528909.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009614.hg.1 ENST00000528909.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009614.hg.1 ENST00000528909.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009614.hg.1 ENST00000528909.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009614.hg.1 ENST00000531340.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009614.hg.1 ENST00000531340.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009614.hg.1 ENST00000531340.5 Ensembl
Affymetrix Human Clariom D Assay TC0100009614.hg.1 NM_000862.2 Refseq
Affymetrix Human Clariom D Assay TC0100009614.hg.1 NM_000862.2 Refseq
Affymetrix Human Clariom D Assay TC0100009614.hg.1 NM_000862.2 Refseq
Affymetrix Human Clariom D Assay TC0100009614.hg.1 NM_000862.2 Refseq
Affymetrix Human Clariom D Assay TC0100009614.hg.1 NM_000862.2 Refseq
Affymetrix Human Clariom D Assay TC0100009614.hg.1 NM_000862.2 Refseq
Affymetrix Human Clariom D Assay TC0100009614.hg.1 NM_000862.2 Refseq
Affymetrix Human Clariom D Assay TC0100009612.hg.1 ENST00000457615.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009612.hg.1 ENST00000457615.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009612.hg.1 ENST00000457615.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009615.hg.1 ENST00000514706.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009615.hg.1 ENST00000514706.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009615.hg.1 ENST00000514706.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100169203.hg.1 ENST00000369413.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100169203.hg.1 ENST00000528909.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100169203.hg.1 NM_000862.2 Refseq
Affymetrix Human Clariom D Assay PSR0100169203.hg.1 ENST00000369413.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100169203.hg.1 ENST00000528909.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100169203.hg.1 NM_000862.2 Refseq
Affymetrix Human Clariom D Assay PSR0100169203.hg.1 TC0100009614.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0100169198.hg.1 ENST00000369413.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100169198.hg.1 ENST00000487520.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100169198.hg.1 ENST00000492140.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100169198.hg.1 ENST00000528909.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100169198.hg.1 ENST00000531340.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100169198.hg.1 NM_000862.2 Refseq
Affymetrix Human Clariom D Assay PSR0100169198.hg.1 ENST00000369413.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100169198.hg.1 ENST00000487520.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100169198.hg.1 ENST00000492140.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100169198.hg.1 ENST00000528909.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100169198.hg.1 ENST00000531340.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100169198.hg.1 NM_000862.2 Refseq
Affymetrix Human Clariom D Assay PSR0100169198.hg.1 TC0100009614.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100009616.hg.1 ENST00000434856.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009616.hg.1 ENST00000434856.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009616.hg.1 ENST00000434856.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100169197.hg.1 ENST00000369413.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100169197.hg.1 ENST00000487520.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100169197.hg.1 ENST00000492140.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100169197.hg.1 ENST00000528909.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100169197.hg.1 ENST00000531340.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100169197.hg.1 NM_000862.2 Refseq
Affymetrix Human Clariom D Assay PSR0100169197.hg.1 ENST00000369413.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100169197.hg.1 ENST00000487520.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100169197.hg.1 ENST00000492140.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100169197.hg.1 ENST00000528909.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100169197.hg.1 ENST00000531340.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100169197.hg.1 NM_000862.2 Refseq
Affymetrix Human Clariom D Assay PSR0100169197.hg.1 TC0100009614.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 197200 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0