This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00429667_gH
See other SV2A GE Assays ›
Gene Symbol:
SV2A
Gene Name:
synaptic vesicle glycoprotein 2A
Gene Aliases:
SV2
Chromosome Location:
Chr.1: 149903318 - 149917882 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00429667_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SV2A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_014849.4 NP_055664.3 4 - 5 1453 77
XM_017002992.1 XP_016858481.1 4 - 5 1239 77
GenBank mRNA AB018279.1 - 4 - 5 1396 77
AK075480.1 - 4 - 5 1413 77
AK291718.1 - 4 - 5 1402 77
BC000776.2 - 3 - 4 548 77
BC045111.1 - 4 - 5 1399 5804662 77
BX537754.1 - 4 - 5 1449 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100015714.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01003195.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SV2A

Gene Ontology Categories:

Function(s) Process(es)

neurotransmitter transport
cellular calcium ion homeostasis
transmembrane transport
receptor activity
protein kinase binding
transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_001278719.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_001278719.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_001278719.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_001278719.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_001278719.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_001278719.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01048947.hg.1 TC01003195.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01048947.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01048947.hg.1 ENST00000369146.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01048947.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_001278719.1 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_001278719.1 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_001278719.1 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_001278719.1 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_001278719.1 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_001278719.1 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay TC0100015714.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay PSR0100217636.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100217636.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100217636.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay PSR0100217636.hg.1 ENST00000369145.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100217636.hg.1 ENST00000369146.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100217636.hg.1 NM_014849.4 Refseq
Affymetrix Human Clariom D Assay PSR0100217636.hg.1 TC0100015714.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0