This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00430664_m1
See other MMP28 GE Assays ›
Gene Symbol:
MMP28
Gene Name:
matrix metallopeptidase 28
Gene Aliases:
EPILYSIN, MM28, MMP-25, MMP-28, MMP25
Chromosome Location:
Chr.17: 35756249 - 35795707 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00430664_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MMP28
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_024302.4 NP_077278.1 7 - 8 1502 141
NR_111988.1 7 - 8 1472 141
XM_011525225.1 XP_011523527.1 6 - 7 1277 141
XM_011525226.2 XP_011523528.1 8 - 9 1614 141
XM_011525227.1 XP_011523529.1 8 - 9 1243 141
XM_011525228.1 XP_011523530.1 8 - 9 1268 141
XM_011525229.2 XP_011523531.1 7 - 8 1143 141
XM_011525232.2 XP_011523534.2 7 - 8 1424 141
XM_017025061.1 XP_016880550.1 8 - 9 1232 141
XM_017025062.1 XP_016880551.1 7 - 8 1498 141
GenBank mRNA AF219624.1 - 7 - 8 1341 141
AF315683.1 - 7 - 8 1187 141
AF330002.1 - 7 - 8 1447 141
AK122604.1 - 7 - 8 1472 141
AY358987.1 - 7 - 8 1377 141

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1700010440.hg.1
This assay targets the same location as HTA transcript cluster(s) TC17001376.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protease 3' Most Homo sapiens 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MMP28

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
negative regulation of macrophage chemotaxis
metalloendopeptidase activity
calcium ion binding
zinc ion binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR17018234.hg.1 ENST00000338839.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17018234.hg.1 NM_024302.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17018234.hg.1 TC17001376.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR17018232.hg.1 ENST00000338839.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17018232.hg.1 NM_024302.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17018232.hg.1 TC17001376.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR17018233.hg.1 ENST00000338839.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17018233.hg.1 NM_024302.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17018233.hg.1 TC17001376.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1700181905.hg.1 NM_024302.4 Refseq
Affymetrix Human Clariom D Assay PSR1700181905.hg.1 NM_032950.3 Refseq
Affymetrix Human Clariom D Assay PSR1700181905.hg.1 NM_024302.4 Refseq
Affymetrix Human Clariom D Assay PSR1700181905.hg.1 NM_032950.3 Refseq
Affymetrix Human Clariom D Assay PSR1700181905.hg.1 TC1700010440.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 196296 - -
ABI Human Genome Survey Microarray Version 2.0 220963 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0