This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00510348_m1
See other RPF1 GE Assays ›
Gene Symbol:
RPF1
Gene Name:
ribosome production factor 1 homolog
Gene Aliases:
BXDC5
Chromosome Location:
Chr.1: 84479237 - 84498350 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00510348_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPF1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025065.6 NP_079341.2 1 - 2 276 94
GenBank mRNA AF322053.1 - 1 - 2 231 94
AK022537.1 - 1 - 2 247 94
BC016051.1 - 1 - 2 238 4853072 94

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100008884.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01000812.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RPF1

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

ribosomal large subunit assembly
maturation of 5.8S rRNA
maturation of LSU-rRNA
RNA binding
protein binding
rRNA primary transcript binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100008884.hg.1 ENST00000370654.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008884.hg.1 ENST00000370654.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008884.hg.1 ENST00000370654.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008884.hg.1 ENST00000370654.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008884.hg.1 ENST00000370654.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008884.hg.1 ENST00000370654.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008884.hg.1 ENST00000370654.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008884.hg.1 ENST00000370654.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008884.hg.1 ENST00000370654.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008884.hg.1 ENST00000370654.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008884.hg.1 ENST00000370654.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008884.hg.1 ENST00000370656.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008884.hg.1 ENST00000370656.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008884.hg.1 ENST00000370656.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008884.hg.1 ENST00000370656.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100008884.hg.1 NM_025065.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008884.hg.1 NM_025065.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008884.hg.1 NM_025065.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008884.hg.1 NM_025065.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008884.hg.1 NM_025065.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008884.hg.1 NM_025065.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008884.hg.1 NM_025065.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008884.hg.1 NM_025065.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008884.hg.1 NM_025065.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008884.hg.1 NM_025065.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008884.hg.1 NM_025065.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100008884.hg.1 NM_025065.6 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01012927.hg.1 TC01000812.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01012927.hg.1 ENST00000370654.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01012927.hg.1 ENST00000370656.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01012927.hg.1 NM_025065.6 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01012926.hg.1 TC01000812.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01012926.hg.1 ENST00000370654.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01012926.hg.1 ENST00000370656.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01012926.hg.1 NM_025065.6 Refseq
Affymetrix Human Clariom D Assay PSR0100163405.hg.1 ENST00000370654.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100163405.hg.1 ENST00000370656.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100163405.hg.1 NM_025065.6 Refseq
Affymetrix Human Clariom D Assay PSR0100163405.hg.1 ENST00000370654.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100163405.hg.1 ENST00000370656.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100163405.hg.1 NM_025065.6 Refseq
Affymetrix Human Clariom D Assay PSR0100163405.hg.1 TC0100008884.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100008884.hg.1 ENST00000370654.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008884.hg.1 ENST00000370654.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008884.hg.1 ENST00000370654.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008884.hg.1 ENST00000370654.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008884.hg.1 ENST00000370654.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008884.hg.1 ENST00000370654.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008884.hg.1 ENST00000370654.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008884.hg.1 ENST00000370654.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008884.hg.1 ENST00000370654.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008884.hg.1 ENST00000370654.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008884.hg.1 ENST00000370654.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008884.hg.1 ENST00000370656.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008884.hg.1 ENST00000370656.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008884.hg.1 ENST00000370656.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008884.hg.1 ENST00000370656.5 Ensembl
Affymetrix Human Clariom D Assay TC0100008884.hg.1 NM_025065.6 Refseq
Affymetrix Human Clariom D Assay TC0100008884.hg.1 NM_025065.6 Refseq
Affymetrix Human Clariom D Assay TC0100008884.hg.1 NM_025065.6 Refseq
Affymetrix Human Clariom D Assay TC0100008884.hg.1 NM_025065.6 Refseq
Affymetrix Human Clariom D Assay TC0100008884.hg.1 NM_025065.6 Refseq
Affymetrix Human Clariom D Assay TC0100008884.hg.1 NM_025065.6 Refseq
Affymetrix Human Clariom D Assay TC0100008884.hg.1 NM_025065.6 Refseq
Affymetrix Human Clariom D Assay TC0100008884.hg.1 NM_025065.6 Refseq
Affymetrix Human Clariom D Assay TC0100008884.hg.1 NM_025065.6 Refseq
Affymetrix Human Clariom D Assay TC0100008884.hg.1 NM_025065.6 Refseq
Affymetrix Human Clariom D Assay TC0100008884.hg.1 NM_025065.6 Refseq
Affymetrix Human Clariom D Assay TC0100008884.hg.1 NM_025065.6 Refseq
Affymetrix Human Clariom D Assay PSR0100163404.hg.1 ENST00000370654.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100163404.hg.1 ENST00000370656.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100163404.hg.1 NM_025065.6 Refseq
Affymetrix Human Clariom D Assay PSR0100163404.hg.1 ENST00000370654.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100163404.hg.1 ENST00000370656.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100163404.hg.1 NM_025065.6 Refseq
Affymetrix Human Clariom D Assay PSR0100163404.hg.1 TC0100008884.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0