This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00535675_g1
See other PPA1 GE Assays ›
Gene Symbol:
PPA1
Gene Name:
pyrophosphatase (inorganic) 1
Gene Aliases:
HEL-S-66p, IOPPP, PP, PP1, SID6-8061
Chromosome Location:
Chr.10: 70202830 - 70233434 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00535675_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021129.3 NP_066952.1 4 - 5 399 81
GenBank mRNA AB026723.1 - 4 - 5 322 81
AF092439.1 - 4 - 5 369 81
AF108211.1 - 4 - 5 280 81
AF119665.1 - 4 - 5 369 81
AF154065.1 - 4 - 5 337 81
AF217186.1 - 4 - 5 292 81
AK058193.1 - 4 - 5 392 81
BC001022.2 - 4 - 5 374 3345496 81
BC061581.1 - 4 - 5 382 6526628 81
BC105034.1 - 4 - 5 365 8144037 81
BC105036.1 - 4 - 5 365 8144039 81
BC107882.1 - 4 - 5 333 6495107 81
BU520836.1 - 4 - 5 380 81
EU794626.1 - 4 - 5 399 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000010914.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10001362.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PPA1

Panther Classification:

Molecular Function -

pyrophosphatase

Gene Ontology Categories:

Function(s) Process(es)

tRNA aminoacylation for protein translation
phosphate-containing compound metabolic process
diphosphate metabolic process
magnesium ion binding
inorganic diphosphatase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373230.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373230.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373230.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373230.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373230.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373230.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373230.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373230.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373230.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373230.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000373232.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000460755.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000460755.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000460755.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000495346.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000495346.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000495346.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000610026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000610026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000610026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000610026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000610026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000610026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000610026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000610026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000610026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000625364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000625364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000625364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000625364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000625364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000625364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000625364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000625364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 ENST00000625364.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000010914.hg.1 NM_021129.3 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 107026 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline