This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00539626_g1
See other ASIC4 GE Assays ›
Gene Symbol:
ASIC4
Gene Name:
acid sensing ion channel subunit family member 4
Gene Aliases:
ACCN4, BNAC4
Chromosome Location:
Chr.2: 219514170 - 219538772 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00539626_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ASIC4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005246671.2 XP_005246728.2 4 - 5 1429 59
XM_017004439.1 XP_016859928.1 4 - 5 588 59
GenBank mRNA AK308804.1 - 4 - 5 1429 59
EL736952.1 - 3 - 4 360 59

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

ion channel

Gene Ontology Categories:

Function(s) Process(es)

transport
sodium ion transmembrane transport
ion channel activity
sodium channel activity
sodium ion transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000347842.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000347842.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000347842.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000347842.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000347842.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000347842.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000347842.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000347842.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000347842.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000347842.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000358078.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000358078.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000358078.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000358078.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000358078.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000358078.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000358078.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000358078.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000358078.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000358078.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000358078.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000461395.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000461395.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000461395.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000461395.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000461395.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000461395.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000461395.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000461395.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000461395.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000461395.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000473709.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000473709.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000473709.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000473709.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000473709.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000473709.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000474489.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000474489.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000474489.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000474489.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000474489.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000474489.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000474489.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000474489.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000474489.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000474489.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000474489.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000474489.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000474489.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 ENST00000474489.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 NM_182847.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 NM_182847.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 NM_182847.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 NM_182847.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 NM_182847.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 NM_182847.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 NM_182847.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 NM_182847.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 NM_182847.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 NM_182847.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200010853.hg.1 NM_182847.2 Refseq
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000347842.7 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000347842.7 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000347842.7 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000347842.7 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000347842.7 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000347842.7 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000347842.7 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000347842.7 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000347842.7 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000347842.7 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000358078.4 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000358078.4 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000358078.4 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000358078.4 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000358078.4 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000358078.4 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000358078.4 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000358078.4 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000358078.4 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000358078.4 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000358078.4 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000461395.5 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000461395.5 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000461395.5 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000461395.5 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000461395.5 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000461395.5 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000461395.5 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000461395.5 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000461395.5 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000461395.5 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000473709.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000473709.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000473709.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000473709.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000473709.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000473709.1 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000474489.5 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000474489.5 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000474489.5 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000474489.5 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000474489.5 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000474489.5 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000474489.5 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000474489.5 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000474489.5 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000474489.5 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000474489.5 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000474489.5 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000474489.5 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 ENST00000474489.5 Ensembl
Affymetrix Human Clariom D Assay TC0200010853.hg.1 NM_182847.2 Refseq
Affymetrix Human Clariom D Assay TC0200010853.hg.1 NM_182847.2 Refseq
Affymetrix Human Clariom D Assay TC0200010853.hg.1 NM_182847.2 Refseq
Affymetrix Human Clariom D Assay TC0200010853.hg.1 NM_182847.2 Refseq
Affymetrix Human Clariom D Assay TC0200010853.hg.1 NM_182847.2 Refseq
Affymetrix Human Clariom D Assay TC0200010853.hg.1 NM_182847.2 Refseq
Affymetrix Human Clariom D Assay TC0200010853.hg.1 NM_182847.2 Refseq
Affymetrix Human Clariom D Assay TC0200010853.hg.1 NM_182847.2 Refseq
Affymetrix Human Clariom D Assay TC0200010853.hg.1 NM_182847.2 Refseq
Affymetrix Human Clariom D Assay TC0200010853.hg.1 NM_182847.2 Refseq
Affymetrix Human Clariom D Assay TC0200010853.hg.1 NM_182847.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0