This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00540876_g1
See other TRMT10B GE Assays ›
Gene Symbol:
TRMT10B
Gene Name:
tRNA methyltransferase 10B
Gene Aliases:
RG9MTD3, bA3J10.9
Chromosome Location:
Chr.9: 37753204 - 37780459 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00540876_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TRMT10B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001286951.1 NP_001273880.1 3 - 4 386 85
NM_144964.3 NP_659401.2 4 - 5 495 85
XM_005251379.2 XP_005251436.1 4 - 5 526 85
XM_011517735.2 XP_011516037.1 4 - 5 517 85
XM_011517736.2 XP_011516038.1 4 - 5 522 85
XM_017014313.1 XP_016869802.1 4 - 5 535 85
GenBank mRNA AK056017.1 - 4 - 5 493 85
AK092718.1 - 3 - 4 376 85
BC057774.1 - 4 - 5 482 30338757 85

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09000217.hg.1

Additional Information:

This assay is in the 5'UTR region of the XM_005251379.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TRMT10B

Panther Classification:

Molecular Function -

RNA methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

methylation
methyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR09002573.hg.1 TC09000217.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR09002573.hg.1 ENST00000297994.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09002573.hg.1 ENST00000377753.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09002573.hg.1 ENST00000377754.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09002573.hg.1 NM_144964.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0