This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00541255_s1
See other IGF1R GE Assays ›
Gene Symbol:
IGF1R
Gene Name:
insulin like growth factor 1 receptor
Gene Aliases:
CD221, IGFIR, IGFR, JTK13
Chromosome Location:
Chr.15: 98648539 - 98964530 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00541255_s1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IGF1R
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000875.4 NP_000866.1 21 11744 79
NM_001291858.1 NP_001278787.1 21 11741 79
XM_017022136.1 XP_016877625.1 21 11247 79
XM_017022137.1 XP_016877626.1 22 10761 79
XM_017022138.1 XP_016877627.1 21 11244 79
GenBank mRNA AB425196.1 - 21 11225 79
BC010607.1 - 1 2755 79
BC088377.1 - 1 775 79
BC111046.1 - 1 1085 79

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene IGF1R

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

immune response
signal transduction
positive regulation of cell proliferation
insulin receptor signaling pathway
phosphatidylinositol 3-kinase signaling
positive regulation of cell migration
peptidyl-tyrosine autophosphorylation
negative regulation of apoptotic process
positive regulation of DNA replication
regulation of JNK cascade
protein autophosphorylation
insulin-like growth factor receptor signaling pathway
phosphatidylinositol-mediated signaling
protein tetramerization
inactivation of MAPKK activity
protein tyrosine kinase activity
insulin-like growth factor-activated receptor activity
insulin receptor binding
protein binding
insulin-like growth factor binding
ATP binding
insulin-like growth factor I binding
identical protein binding
phosphatidylinositol 3-kinase binding
insulin binding
insulin receptor substrate binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline