This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00542762_m1
See other UNC45B GE Assays ›
Gene Symbol:
UNC45B
Gene Name:
unc-45 myosin chaperone B
Gene Aliases:
CMYA4, CTRCT43, SMUNC45, UNC-45B, UNC45
Chromosome Location:
Chr.17: 35147793 - 35189345 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00542762_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UNC45B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_173167.3 NP_775259.1 11 - 12 1689 61
GenBank mRNA AF539794.1 - 11 - 12 1689 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1700012235.hg.1
This assay targets the same location as HTA transcript cluster(s) TC17000395.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene UNC45B

Gene Ontology Categories:

Function(s) Process(es)

lens development in camera-type eye
muscle organ development
cell differentiation
chaperone-mediated protein folding
protein binding
Hsp90 protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR17004973.hg.1 ENST00000268876.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17004973.hg.1 ENST00000378449.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17004973.hg.1 ENST00000394570.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17004973.hg.1 ENST00000433649.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17004973.hg.1 NM_001033576.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17004973.hg.1 NM_173167.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17004973.hg.1 TC17000395.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1700201916.hg.1 ENST00000268876.9 Ensembl
Affymetrix Human Clariom D Assay PSR1700201916.hg.1 ENST00000394570.6 Ensembl
Affymetrix Human Clariom D Assay PSR1700201916.hg.1 ENST00000591048.2 Ensembl
Affymetrix Human Clariom D Assay PSR1700201916.hg.1 NM_001033576.1 Refseq
Affymetrix Human Clariom D Assay PSR1700201916.hg.1 NM_001267052.1 Refseq
Affymetrix Human Clariom D Assay PSR1700201916.hg.1 NM_001308281.1 Refseq
Affymetrix Human Clariom D Assay PSR1700201916.hg.1 NM_173167.3 Refseq
Affymetrix Human Clariom D Assay PSR1700201916.hg.1 ENST00000268876.9 Ensembl
Affymetrix Human Clariom D Assay PSR1700201916.hg.1 ENST00000394570.6 Ensembl
Affymetrix Human Clariom D Assay PSR1700201916.hg.1 ENST00000591048.2 Ensembl
Affymetrix Human Clariom D Assay PSR1700201916.hg.1 NM_001033576.1 Refseq
Affymetrix Human Clariom D Assay PSR1700201916.hg.1 NM_001267052.1 Refseq
Affymetrix Human Clariom D Assay PSR1700201916.hg.1 NM_001308281.1 Refseq
Affymetrix Human Clariom D Assay PSR1700201916.hg.1 NM_173167.3 Refseq
Affymetrix Human Clariom D Assay PSR1700201916.hg.1 TC1700012235.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0