This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00543759_m1
See other P2RX3 GE Assays ›
Gene Symbol:
P2RX3
Gene Name:
purinergic receptor P2X 3
Gene Aliases:
P2X3
Chromosome Location:
Chr.11: 57335943 - 57372399 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00543759_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: P2RX3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002559.3 NP_002550.2 4 - 5 576 105
XM_011545069.2 XP_011543371.1 5 - 6 1237 105
XM_011545070.2 XP_011543372.1 5 - 6 543 105
XM_011545072.2 XP_011543374.1 5 - 6 1369 105
GenBank mRNA AB016608.1 - 4 - 5 392 105
BC069162.1 - 4 - 5 468 7216977 105
BC074792.2 - 4 - 5 423 30915565 105
BC074793.2 - 4 - 5 423 30915348 105
BC109287.2 - 4 - 5 426 40031972 105

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100007670.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11000461.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene P2RX3

Panther Classification:

Molecular Function -

ligand-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
transport
signal transduction
neuromuscular synaptic transmission
blood coagulation
response to heat
response to cold
response to mechanical stimulus
response to carbohydrate
positive regulation of calcium ion transport into cytosol
urinary bladder smooth muscle contraction
neuronal action potential
peristalsis
response to ATP
purinergic nucleotide receptor signaling pathway
regulation of synaptic plasticity
positive regulation of calcium-mediated signaling
sensory perception of taste
protein homooligomerization
behavioral response to formalin induced pain
cation transmembrane transport
positive regulation of sensory perception of pain
purinergic nucleotide receptor activity
extracellular ATP-gated cation channel activity
ATP binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000533436.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000533436.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000533436.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000533436.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000533436.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000534820.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000534820.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000534820.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000534820.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000534820.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000534820.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11005579.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11005579.hg.1 ENST00000439993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11005579.hg.1 NM_002559.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11005579.hg.1 TC11000461.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1100153942.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100153942.hg.1 ENST00000534820.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100153942.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100153942.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay PSR1100153942.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100153942.hg.1 ENST00000534820.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100153942.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100153942.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay PSR1100153942.hg.1 TC1100007670.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000263314.2 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000533436.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000533436.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000533436.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000533436.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000533436.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000534820.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000534820.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000534820.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000534820.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000534820.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000534820.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 ENST00000616487.4 Ensembl
Affymetrix Human Clariom D Assay TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay TC1100007670.hg.1 NM_002559.4 Refseq
Affymetrix Human Clariom D Assay TC1100007670.hg.1 NM_002559.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0