This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00545356_m1
See other PDILT GE Assays ›
Gene Symbol:
PDILT
Gene Name:
protein disulfide isomerase like, testis expressed
Gene Aliases:
PDIA7
Chromosome Location:
Chr.16: 20359170 - 20404742 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00545356_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PDILT
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_174924.1 NP_777584.1 10 - 11 1634 77
XM_011545766.2 XP_011544068.1 6 - 7 819 77
GenBank mRNA AK097476.1 - 10 - 11 1660 77
BC042607.1 - 10 - 11 1634 5169707 77
BC044936.1 - 10 - 11 1634 5169707 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600009656.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16000925.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PDILT

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein folding
multicellular organism development
spermatid development
cell migration
response to endoplasmic reticulum stress
cell redox homeostasis
protein disulfide isomerase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR16013278.hg.1 ENST00000302451.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16013278.hg.1 NM_174924.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16013278.hg.1 TC16000925.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1600009656.hg.1 ENST00000302451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009656.hg.1 ENST00000302451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009656.hg.1 ENST00000302451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009656.hg.1 ENST00000302451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009656.hg.1 ENST00000302451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009656.hg.1 ENST00000302451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009656.hg.1 ENST00000302451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009656.hg.1 ENST00000302451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009656.hg.1 ENST00000302451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009656.hg.1 ENST00000302451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009656.hg.1 ENST00000302451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009656.hg.1 ENST00000302451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009656.hg.1 ENST00000302451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009656.hg.1 ENST00000302451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009656.hg.1 ENST00000302451.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009656.hg.1 ENST00000575561.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009656.hg.1 ENST00000575561.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009656.hg.1 ENST00000575561.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009656.hg.1 ENST00000575561.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009656.hg.1 NM_174924.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009656.hg.1 NM_174924.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009656.hg.1 NM_174924.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009656.hg.1 NM_174924.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009656.hg.1 NM_174924.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009656.hg.1 NM_174924.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009656.hg.1 NM_174924.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009656.hg.1 NM_174924.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009656.hg.1 NM_174924.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009656.hg.1 NM_174924.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009656.hg.1 NM_174924.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009656.hg.1 NM_174924.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009656.hg.1 NM_174924.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009656.hg.1 NM_174924.1 Refseq
Affymetrix Human Clariom D Assay TC1600009656.hg.1 ENST00000302451.8 Ensembl
Affymetrix Human Clariom D Assay TC1600009656.hg.1 ENST00000302451.8 Ensembl
Affymetrix Human Clariom D Assay TC1600009656.hg.1 ENST00000302451.8 Ensembl
Affymetrix Human Clariom D Assay TC1600009656.hg.1 ENST00000302451.8 Ensembl
Affymetrix Human Clariom D Assay TC1600009656.hg.1 ENST00000302451.8 Ensembl
Affymetrix Human Clariom D Assay TC1600009656.hg.1 ENST00000302451.8 Ensembl
Affymetrix Human Clariom D Assay TC1600009656.hg.1 ENST00000302451.8 Ensembl
Affymetrix Human Clariom D Assay TC1600009656.hg.1 ENST00000302451.8 Ensembl
Affymetrix Human Clariom D Assay TC1600009656.hg.1 ENST00000302451.8 Ensembl
Affymetrix Human Clariom D Assay TC1600009656.hg.1 ENST00000302451.8 Ensembl
Affymetrix Human Clariom D Assay TC1600009656.hg.1 ENST00000302451.8 Ensembl
Affymetrix Human Clariom D Assay TC1600009656.hg.1 ENST00000302451.8 Ensembl
Affymetrix Human Clariom D Assay TC1600009656.hg.1 ENST00000302451.8 Ensembl
Affymetrix Human Clariom D Assay TC1600009656.hg.1 ENST00000302451.8 Ensembl
Affymetrix Human Clariom D Assay TC1600009656.hg.1 ENST00000302451.8 Ensembl
Affymetrix Human Clariom D Assay TC1600009656.hg.1 ENST00000575561.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009656.hg.1 ENST00000575561.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009656.hg.1 ENST00000575561.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009656.hg.1 ENST00000575561.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009656.hg.1 NM_174924.1 Refseq
Affymetrix Human Clariom D Assay TC1600009656.hg.1 NM_174924.1 Refseq
Affymetrix Human Clariom D Assay TC1600009656.hg.1 NM_174924.1 Refseq
Affymetrix Human Clariom D Assay TC1600009656.hg.1 NM_174924.1 Refseq
Affymetrix Human Clariom D Assay TC1600009656.hg.1 NM_174924.1 Refseq
Affymetrix Human Clariom D Assay TC1600009656.hg.1 NM_174924.1 Refseq
Affymetrix Human Clariom D Assay TC1600009656.hg.1 NM_174924.1 Refseq
Affymetrix Human Clariom D Assay TC1600009656.hg.1 NM_174924.1 Refseq
Affymetrix Human Clariom D Assay TC1600009656.hg.1 NM_174924.1 Refseq
Affymetrix Human Clariom D Assay TC1600009656.hg.1 NM_174924.1 Refseq
Affymetrix Human Clariom D Assay TC1600009656.hg.1 NM_174924.1 Refseq
Affymetrix Human Clariom D Assay TC1600009656.hg.1 NM_174924.1 Refseq
Affymetrix Human Clariom D Assay TC1600009656.hg.1 NM_174924.1 Refseq
Affymetrix Human Clariom D Assay TC1600009656.hg.1 NM_174924.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173939.hg.1 ENST00000302451.8 Ensembl
Affymetrix Human Clariom D Assay PSR1600173939.hg.1 NM_174924.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173939.hg.1 TC1600009656.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1600173939.hg.1 ENST00000302451.8 Ensembl
Affymetrix Human Clariom D Assay PSR1600173939.hg.1 NM_174924.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline