This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00559734_m1
See other GPX5 GE Assays ›
Gene Symbol:
GPX5
Gene Name:
glutathione peroxidase 5
Gene Aliases:
HEL-S-75p
Chromosome Location:
Chr.6: 28526012 - 28534951 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00559734_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPX5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003996.3 NP_003987.2 2 - 3 243 108
GenBank mRNA AK093099.1 - 2 - 3 439 108
BC128159.1 - 2 - 3 243 108
BC128160.1 - 2 - 3 243 108

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600007423.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06000286.hg.1, TC6_cox_hap2000001.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GPX5

Panther Classification:

Molecular Function -

peroxidase

Gene Ontology Categories:

Function(s) Process(es)

response to reactive oxygen species
lipid metabolic process
cellular response to oxidative stress
oxidation-reduction process
cellular oxidant detoxification
glutathione peroxidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2000005.hg.1 TC6_cox_hap2000001.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2000005.hg.1 NM_001509.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2000005.hg.1 NM_003996.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 ENST00000412168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 ENST00000412168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 ENST00000412168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 ENST00000412168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 ENST00000412168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 ENST00000412168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 ENST00000412168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 ENST00000412168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 ENST00000412168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 ENST00000442674.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 ENST00000442674.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 ENST00000442674.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 ENST00000442674.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 ENST00000442674.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 ENST00000442674.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 ENST00000442674.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 ENST00000442674.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 ENST00000442674.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 ENST00000442674.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 ENST00000469384.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 ENST00000469384.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 ENST00000469384.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 ENST00000469384.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 ENST00000469384.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 ENST00000483784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 ENST00000483784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 ENST00000483784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 ENST00000483784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 NM_001509.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 NM_001509.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 NM_001509.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 NM_001509.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 NM_001509.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 NM_001509.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 NM_001509.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 NM_001509.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 NM_003996.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 NM_003996.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 NM_003996.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 NM_003996.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 NM_003996.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 NM_003996.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600007423.hg.1 NM_003996.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06002388.hg.1 TC06000286.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR06002388.hg.1 ENST00000412168.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06002388.hg.1 ENST00000442674.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06002388.hg.1 ENST00000469384.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06002388.hg.1 NM_001509.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06002388.hg.1 NM_003996.3 Refseq
Affymetrix Human Clariom D Assay TC0600007423.hg.1 ENST00000412168.6 Ensembl
Affymetrix Human Clariom D Assay TC0600007423.hg.1 ENST00000412168.6 Ensembl
Affymetrix Human Clariom D Assay TC0600007423.hg.1 ENST00000412168.6 Ensembl
Affymetrix Human Clariom D Assay TC0600007423.hg.1 ENST00000412168.6 Ensembl
Affymetrix Human Clariom D Assay TC0600007423.hg.1 ENST00000412168.6 Ensembl
Affymetrix Human Clariom D Assay TC0600007423.hg.1 ENST00000412168.6 Ensembl
Affymetrix Human Clariom D Assay TC0600007423.hg.1 ENST00000412168.6 Ensembl
Affymetrix Human Clariom D Assay TC0600007423.hg.1 ENST00000412168.6 Ensembl
Affymetrix Human Clariom D Assay TC0600007423.hg.1 ENST00000412168.6 Ensembl
Affymetrix Human Clariom D Assay TC0600007423.hg.1 ENST00000442674.6 Ensembl
Affymetrix Human Clariom D Assay TC0600007423.hg.1 ENST00000442674.6 Ensembl
Affymetrix Human Clariom D Assay TC0600007423.hg.1 ENST00000442674.6 Ensembl
Affymetrix Human Clariom D Assay TC0600007423.hg.1 ENST00000442674.6 Ensembl
Affymetrix Human Clariom D Assay TC0600007423.hg.1 ENST00000442674.6 Ensembl
Affymetrix Human Clariom D Assay TC0600007423.hg.1 ENST00000442674.6 Ensembl
Affymetrix Human Clariom D Assay TC0600007423.hg.1 ENST00000442674.6 Ensembl
Affymetrix Human Clariom D Assay TC0600007423.hg.1 ENST00000442674.6 Ensembl
Affymetrix Human Clariom D Assay TC0600007423.hg.1 ENST00000442674.6 Ensembl
Affymetrix Human Clariom D Assay TC0600007423.hg.1 ENST00000442674.6 Ensembl
Affymetrix Human Clariom D Assay TC0600007423.hg.1 ENST00000469384.1 Ensembl
Affymetrix Human Clariom D Assay TC0600007423.hg.1 ENST00000469384.1 Ensembl
Affymetrix Human Clariom D Assay TC0600007423.hg.1 ENST00000469384.1 Ensembl
Affymetrix Human Clariom D Assay TC0600007423.hg.1 ENST00000469384.1 Ensembl
Affymetrix Human Clariom D Assay TC0600007423.hg.1 ENST00000469384.1 Ensembl
Affymetrix Human Clariom D Assay TC0600007423.hg.1 ENST00000483784.1 Ensembl
Affymetrix Human Clariom D Assay TC0600007423.hg.1 ENST00000483784.1 Ensembl
Affymetrix Human Clariom D Assay TC0600007423.hg.1 ENST00000483784.1 Ensembl
Affymetrix Human Clariom D Assay TC0600007423.hg.1 ENST00000483784.1 Ensembl
Affymetrix Human Clariom D Assay TC0600007423.hg.1 NM_001509.2 Refseq
Affymetrix Human Clariom D Assay TC0600007423.hg.1 NM_001509.2 Refseq
Affymetrix Human Clariom D Assay TC0600007423.hg.1 NM_001509.2 Refseq
Affymetrix Human Clariom D Assay TC0600007423.hg.1 NM_001509.2 Refseq
Affymetrix Human Clariom D Assay TC0600007423.hg.1 NM_001509.2 Refseq
Affymetrix Human Clariom D Assay TC0600007423.hg.1 NM_001509.2 Refseq
Affymetrix Human Clariom D Assay TC0600007423.hg.1 NM_001509.2 Refseq
Affymetrix Human Clariom D Assay TC0600007423.hg.1 NM_001509.2 Refseq
Affymetrix Human Clariom D Assay TC0600007423.hg.1 NM_003996.3 Refseq
Affymetrix Human Clariom D Assay TC0600007423.hg.1 NM_003996.3 Refseq
Affymetrix Human Clariom D Assay TC0600007423.hg.1 NM_003996.3 Refseq
Affymetrix Human Clariom D Assay TC0600007423.hg.1 NM_003996.3 Refseq
Affymetrix Human Clariom D Assay TC0600007423.hg.1 NM_003996.3 Refseq
Affymetrix Human Clariom D Assay TC0600007423.hg.1 NM_003996.3 Refseq
Affymetrix Human Clariom D Assay TC0600007423.hg.1 NM_003996.3 Refseq
Affymetrix Human Clariom D Assay PSR0600150280.hg.1 ENST00000412168.6 Ensembl
Affymetrix Human Clariom D Assay PSR0600150280.hg.1 ENST00000442674.6 Ensembl
Affymetrix Human Clariom D Assay PSR0600150280.hg.1 ENST00000469384.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600150280.hg.1 ENST00000483784.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600150280.hg.1 NM_001509.2 Refseq
Affymetrix Human Clariom D Assay PSR0600150280.hg.1 NM_003996.3 Refseq
Affymetrix Human Clariom D Assay PSR0600150280.hg.1 ENST00000412168.6 Ensembl
Affymetrix Human Clariom D Assay PSR0600150280.hg.1 ENST00000442674.6 Ensembl
Affymetrix Human Clariom D Assay PSR0600150280.hg.1 ENST00000469384.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600150280.hg.1 ENST00000483784.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600150280.hg.1 NM_001509.2 Refseq
Affymetrix Human Clariom D Assay PSR0600150280.hg.1 NM_003996.3 Refseq
Affymetrix Human Clariom D Assay PSR0600150280.hg.1 TC0600007423.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0