This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00602143_m1
See other NDUFS5 GE Assays ›
Gene Symbol:
NDUFS5
Gene Name:
NADH:ubiquinone oxidoreductase subunit S5
Gene Aliases:
CI-15k, CI15K
Chromosome Location:
Chr.1: 39026295 - 39034636 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00602143_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NDUFS5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_004552.2 NP_004543.1 1 - 2 107 126
GenBank mRNA AF047434.1 - 1 - 2 68 126
AK312188.1 - 1 - 2 81 126

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100007886.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01000492.hg.1

Additional Information:

This assay is in the 5'UTR region of the AF047434.1,AK312188.1,NM_004552.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NDUFS5

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

mitochondrial electron transport, NADH to ubiquinone
mitochondrial respiratory chain complex I assembly
NADH dehydrogenase (ubiquinone) activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100008330.hg.1 ENST00000450668.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01007520.hg.1 TC01000492.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01007520.hg.1 ENST00000372969.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01007520.hg.1 NM_004552.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015016.hg.1 ENST00000484004.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011397.hg.1 ENST00000412983.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007886.hg.1 ENST00000372967.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007886.hg.1 ENST00000372967.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007886.hg.1 ENST00000372967.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007886.hg.1 ENST00000372967.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007886.hg.1 ENST00000372969.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007886.hg.1 ENST00000372969.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007886.hg.1 ENST00000372969.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007886.hg.1 ENST00000372969.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007886.hg.1 NM_001184979.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007886.hg.1 NM_001184979.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007886.hg.1 NM_001184979.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007886.hg.1 NM_001184979.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007886.hg.1 NM_004552.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007886.hg.1 NM_004552.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007886.hg.1 NM_004552.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007886.hg.1 NM_004552.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011687.hg.1 ENST00000437256.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010227.hg.1 ENST00000580924.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01007516.hg.1 TC01000492.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01007516.hg.1 ENST00000372967.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01007516.hg.1 ENST00000372969.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01007516.hg.1 NM_001184979.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01007516.hg.1 NM_004552.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01007521.hg.1 TC01000492.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01007521.hg.1 ENST00000372967.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01007521.hg.1 ENST00000372969.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01007521.hg.1 NM_001184979.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01007521.hg.1 NM_004552.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01007522.hg.1 TC01000492.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01007522.hg.1 ENST00000372967.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01007522.hg.1 ENST00000372969.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01007522.hg.1 NM_001184979.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01007522.hg.1 NM_004552.2 Refseq
Affymetrix Human Clariom D Assay TC0400011397.hg.1 ENST00000412983.2 Ensembl
Affymetrix Human Clariom D Assay PSR0100155443.hg.1 ENST00000372967.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100155443.hg.1 ENST00000372969.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100155443.hg.1 NM_001184979.1 Refseq
Affymetrix Human Clariom D Assay PSR0100155443.hg.1 NM_004552.2 Refseq
Affymetrix Human Clariom D Assay PSR0100155443.hg.1 ENST00000372967.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100155443.hg.1 ENST00000372969.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100155443.hg.1 NM_001184979.1 Refseq
Affymetrix Human Clariom D Assay PSR0100155443.hg.1 NM_004552.2 Refseq
Affymetrix Human Clariom D Assay PSR0100155443.hg.1 TC0100007886.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100008330.hg.1 ENST00000450668.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010227.hg.1 ENST00000580924.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007886.hg.1 ENST00000372967.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007886.hg.1 ENST00000372967.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007886.hg.1 ENST00000372967.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007886.hg.1 ENST00000372967.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007886.hg.1 ENST00000372969.7 Ensembl
Affymetrix Human Clariom D Assay TC0100007886.hg.1 ENST00000372969.7 Ensembl
Affymetrix Human Clariom D Assay TC0100007886.hg.1 ENST00000372969.7 Ensembl
Affymetrix Human Clariom D Assay TC0100007886.hg.1 ENST00000372969.7 Ensembl
Affymetrix Human Clariom D Assay TC0100007886.hg.1 NM_001184979.1 Refseq
Affymetrix Human Clariom D Assay TC0100007886.hg.1 NM_001184979.1 Refseq
Affymetrix Human Clariom D Assay TC0100007886.hg.1 NM_001184979.1 Refseq
Affymetrix Human Clariom D Assay TC0100007886.hg.1 NM_001184979.1 Refseq
Affymetrix Human Clariom D Assay TC0100007886.hg.1 NM_004552.2 Refseq
Affymetrix Human Clariom D Assay TC0100007886.hg.1 NM_004552.2 Refseq
Affymetrix Human Clariom D Assay TC0100007886.hg.1 NM_004552.2 Refseq
Affymetrix Human Clariom D Assay TC0100007886.hg.1 NM_004552.2 Refseq
Affymetrix Human Clariom D Assay TC0400011687.hg.1 ENST00000437256.2 Ensembl
Affymetrix Human Clariom D Assay PSR0100155446.hg.1 ENST00000372967.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100155446.hg.1 ENST00000372969.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100155446.hg.1 NM_001184979.1 Refseq
Affymetrix Human Clariom D Assay PSR0100155446.hg.1 NM_004552.2 Refseq
Affymetrix Human Clariom D Assay PSR0100155446.hg.1 ENST00000372967.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100155446.hg.1 ENST00000372969.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100155446.hg.1 NM_001184979.1 Refseq
Affymetrix Human Clariom D Assay PSR0100155446.hg.1 NM_004552.2 Refseq
Affymetrix Human Clariom D Assay PSR0100155446.hg.1 TC0100007886.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100015016.hg.1 ENST00000484004.2 Ensembl
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 133148 - -
ABI Human Genome Survey Microarray Version 2.0 140600 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0