This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00604460_g1
See other IGDCC3 GE Assays ›
Gene Symbol:
IGDCC3
Gene Name:
immunoglobulin superfamily DCC subclass member 3
Gene Aliases:
HsT18880, PUNC
Chromosome Location:
Chr.15: 65327127 - 65378040 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00604460_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IGDCC3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_004884.3 NP_004875.2 7 - 8 1398 59
XM_011522241.1 XP_011520543.1 7 - 8 1340 59
XM_011522243.1 XP_011520545.1 6 - 7 924 59
XM_011522244.1 XP_011520546.1 6 - 7 913 59
XM_011522245.2 XP_011520547.1 7 - 8 1340 59
GenBank mRNA AK095529.1 - 6 - 7 913 59
BC042054.1 - 7 - 8 1346 5579570 59
BC067107.1 - 7 - 8 1398 6644528 59

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1500009795.hg.1
This assay targets the same location as HTA transcript cluster(s) TC15001565.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene IGDCC3

Gene Ontology Categories:

Function(s)

neuromuscular process controlling balance

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000558354.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000558354.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000558354.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000558354.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000558354.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000558354.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000558354.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000558354.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000558354.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000558354.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000558354.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000558354.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000558354.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000559058.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000559058.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000559058.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000559058.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000559231.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000559231.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000559231.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000559231.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000559231.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000559947.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 ENST00000559947.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR15014948.hg.1 ENST00000327987.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15014948.hg.1 ENST00000443278.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15014948.hg.1 ENST00000559231.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15014948.hg.1 NM_004884.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR15014948.hg.1 TC15001565.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000558354.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000558354.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000558354.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000558354.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000558354.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000558354.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000558354.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000558354.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000558354.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000558354.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000558354.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000558354.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000558354.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000559058.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000559058.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000559058.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000559058.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000559231.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000559231.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000559231.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000559231.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000559231.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000559947.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 ENST00000559947.1 Ensembl
Affymetrix Human Clariom D Assay TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Clariom D Assay TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Clariom D Assay TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Clariom D Assay TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Clariom D Assay TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Clariom D Assay TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Clariom D Assay TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Clariom D Assay TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Clariom D Assay TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Clariom D Assay TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Clariom D Assay TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Clariom D Assay TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Clariom D Assay TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Clariom D Assay TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Clariom D Assay TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Clariom D Assay TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Clariom D Assay TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Clariom D Assay TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Clariom D Assay TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Clariom D Assay TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Clariom D Assay TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Clariom D Assay TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Clariom D Assay TC1500009795.hg.1 NM_004884.3 Refseq
Affymetrix Human Clariom D Assay PSR1500172389.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Clariom D Assay PSR1500172389.hg.1 ENST00000558354.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500172389.hg.1 ENST00000559231.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500172389.hg.1 NM_004884.3 Refseq
Affymetrix Human Clariom D Assay PSR1500172389.hg.1 ENST00000327987.8 Ensembl
Affymetrix Human Clariom D Assay PSR1500172389.hg.1 ENST00000558354.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500172389.hg.1 ENST00000559231.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500172389.hg.1 NM_004884.3 Refseq
Affymetrix Human Clariom D Assay PSR1500172389.hg.1 TC1500009795.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0