This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00606481_m1
See other SSR3 GE Assays ›
Gene Symbol:
SSR3
Gene Name:
signal sequence receptor subunit 3
Gene Aliases:
TRAPG
Chromosome Location:
Chr.3: 156539553 - 156555200 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00606481_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SSR3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001308197.1 NP_001295126.1 1 - 2 244 89
NM_007107.4 NP_009038.1 1 - 2 244 89
GenBank mRNA AF087907.1 - 1 - 2 192 89
AF110647.1 - 1 - 2 189 89
AK304364.1 - 1 - 2 142 89
AK312935.1 - 1 - 2 151 89
BC017203.1 - 1 - 2 190 3618589 89
BT006669.1 - 1 - 2 133 89
DA135338.1 - 1 - 2 228 89
DA750309.1 - 1 - 2 228 89
HY035536.1 - 1 - 2 244 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300012897.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03001933.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SSR3

Gene Ontology Categories:

Function(s)

SRP-dependent cotranslational protein targeting to membrane

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100008809.hg.1 ENST00000448345.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000265044.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000265044.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000265044.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000265044.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000265044.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000265044.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000265044.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000265044.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000265044.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000265044.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000265044.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000265044.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000265044.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000463503.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000463503.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000463503.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000463503.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000463503.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000463503.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000464138.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000464138.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000464138.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000464138.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000464138.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000464138.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000464138.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000467733.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000467733.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000467733.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000467733.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000467733.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000467733.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000467789.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000467789.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000467789.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000467789.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000467789.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000467789.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000467789.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000467789.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000467789.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000467789.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000467789.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000476217.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000476217.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000476217.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000476217.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000476217.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000476217.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000476217.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000476217.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000476217.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000478842.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000478842.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000478842.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000478842.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000478842.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000478842.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000478842.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000496050.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000496050.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000496050.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000496050.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000496050.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000498205.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000498205.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000498205.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000498205.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000498205.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000498205.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 ENST00000498205.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308197.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308197.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308197.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308197.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308197.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308197.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308197.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308197.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308197.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308197.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308197.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308197.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308197.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308197.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308197.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308204.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_001308205.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_007107.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_007107.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_007107.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_007107.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_007107.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_007107.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_007107.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_007107.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_007107.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_007107.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_007107.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_007107.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_007107.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012897.hg.1 NM_007107.4 Refseq
Affymetrix Human Clariom D Assay TC0100008809.hg.1 ENST00000448345.2 Ensembl
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 124871 - -
ABI Human Genome Survey Microarray Version 2.0 215985 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x8dcn:80/100.66.79.134:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0