This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00608418_m1
See other POLR2H GE Assays ›
Gene Symbol:
POLR2H
Gene Name:
RNA polymerase II subunit H
Gene Aliases:
RPABC3, RPB17, RPB8
Chromosome Location:
Chr.3: 184361710 - 184368595 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00608418_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: POLR2H
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001278698.1 NP_001265627.1 3 - 4 762 86
NM_001278699.1 NP_001265628.1 3 - 4 771 86
NM_001278700.1 NP_001265629.1 3 - 4 612 86
NM_006232.3 NP_006223.2 3 - 4 762 86
XM_006713666.2 XP_006713729.1 4 - 5 1300 86
XM_006713667.2 XP_006713730.1 4 - 5 1049 86
XM_006713668.3 XP_006713731.1 4 - 5 1309 86
XM_006713670.2 XP_006713733.1 4 - 5 1309 86
GenBank mRNA BC000739.2 - 3 - 4 312 3504683 86
BG529732.1 - 3 - 4 343 86
BM541682.1 - 3 - 4 569 86
U37689.1 - 3 - 4 417 86
Z49199.1 - 3 - 4 330 86

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene POLR2H

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

mRNA splicing, via spliceosome
transcription-coupled nucleotide-excision repair
transcription initiation from RNA polymerase I promoter
transcription elongation from RNA polymerase I promoter
termination of RNA polymerase I transcription
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
7-methylguanosine mRNA capping
transcription from RNA polymerase III promoter
fibroblast growth factor receptor signaling pathway
gene expression
gene silencing by RNA
positive regulation of type I interferon production
somatic stem cell population maintenance
snRNA transcription from RNA polymerase II promoter
positive regulation of gene expression, epigenetic
positive regulation of viral transcription
RNA polymerase I activity
RNA polymerase II activity
RNA polymerase III activity
DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0