This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00609161_m1
See other EXT1 GE Assays ›
Gene Symbol:
EXT1
Gene Name:
exostosin glycosyltransferase 1
Gene Aliases:
EXT, LGCR, LGS, TRPS2, TTV
Chromosome Location:
Chr.8: 117797496 - 118111819 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00609161_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EXT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000127.2 NP_000118.2 7 - 8 2405 65
GenBank mRNA BC001174.1 - 7 - 8 1798 3502232 65
S79639.1 - 7 - 8 2283 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0800011579.hg.1
This assay targets the same location as HTA transcript cluster(s) TC08001548.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EXT1

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s) Process(es)

skeletal system development
ossification
glycosaminoglycan biosynthetic process
protein glycosylation
signal transduction
gastrulation
axon guidance
endoderm development
mesoderm development
heparan sulfate proteoglycan biosynthetic process
heparan sulfate proteoglycan biosynthetic process, polysaccharide chain biosynthetic process
olfactory bulb development
cellular polysaccharide biosynthetic process
embryonic skeletal joint development
acetylglucosaminyltransferase activity
glucuronosyltransferase activity
transferase activity, transferring glycosyl groups
heparan sulfate N-acetylglucosaminyltransferase activity
protein homodimerization activity
metal ion binding
protein heterodimerization activity
glucuronosyl-N-acetylglucosaminyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase activity
N-acetylglucosaminyl-proteoglycan 4-beta-glucuronosyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR08020395.hg.1 TC08001548.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR08020395.hg.1 ENST00000378204.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08020395.hg.1 NM_000127.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000436216.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000436216.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000436216.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000436216.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000436216.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000437196.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000437196.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000437196.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000437196.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000437196.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000437196.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000437196.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000437196.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000437196.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000437196.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000437196.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 ENST00000437196.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 NM_000127.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 NM_000127.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 NM_000127.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 NM_000127.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 NM_000127.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 NM_000127.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 NM_000127.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 NM_000127.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 NM_000127.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 NM_000127.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 NM_000127.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 NM_000127.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 NM_000127.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 NM_000127.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 NM_000127.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 NM_000127.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 NM_000127.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011579.hg.1 NM_000127.2 Refseq
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000436216.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000436216.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000436216.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000436216.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000436216.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000437196.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000437196.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000437196.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000437196.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000437196.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000437196.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000437196.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000437196.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000437196.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000437196.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000437196.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 ENST00000437196.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011579.hg.1 NM_000127.2 Refseq
Affymetrix Human Clariom D Assay TC0800011579.hg.1 NM_000127.2 Refseq
Affymetrix Human Clariom D Assay TC0800011579.hg.1 NM_000127.2 Refseq
Affymetrix Human Clariom D Assay TC0800011579.hg.1 NM_000127.2 Refseq
Affymetrix Human Clariom D Assay TC0800011579.hg.1 NM_000127.2 Refseq
Affymetrix Human Clariom D Assay TC0800011579.hg.1 NM_000127.2 Refseq
Affymetrix Human Clariom D Assay TC0800011579.hg.1 NM_000127.2 Refseq
Affymetrix Human Clariom D Assay TC0800011579.hg.1 NM_000127.2 Refseq
Affymetrix Human Clariom D Assay TC0800011579.hg.1 NM_000127.2 Refseq
Affymetrix Human Clariom D Assay TC0800011579.hg.1 NM_000127.2 Refseq
Affymetrix Human Clariom D Assay TC0800011579.hg.1 NM_000127.2 Refseq
Affymetrix Human Clariom D Assay TC0800011579.hg.1 NM_000127.2 Refseq
Affymetrix Human Clariom D Assay TC0800011579.hg.1 NM_000127.2 Refseq
Affymetrix Human Clariom D Assay TC0800011579.hg.1 NM_000127.2 Refseq
Affymetrix Human Clariom D Assay TC0800011579.hg.1 NM_000127.2 Refseq
Affymetrix Human Clariom D Assay TC0800011579.hg.1 NM_000127.2 Refseq
Affymetrix Human Clariom D Assay TC0800011579.hg.1 NM_000127.2 Refseq
Affymetrix Human Clariom D Assay TC0800011579.hg.1 NM_000127.2 Refseq
Affymetrix Human Clariom D Assay PSR0800178673.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Clariom D Assay PSR0800178673.hg.1 ENST00000437196.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800178673.hg.1 NM_000127.2 Refseq
Affymetrix Human Clariom D Assay PSR0800178673.hg.1 ENST00000378204.6 Ensembl
Affymetrix Human Clariom D Assay PSR0800178673.hg.1 ENST00000437196.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800178673.hg.1 NM_000127.2 Refseq
Affymetrix Human Clariom D Assay PSR0800178673.hg.1 TC0800011579.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0