This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00609432_g1
See other TAP1 GE Assays ›
Gene Symbol:
TAP1
Gene Name:
transporter 1, ATP binding cassette subfamily B member
Gene Aliases:
ABC17, ABCB2, APT1, D6S114E, PSF-1, PSF1, RING4, TAP1*0102N, TAP1N
Chromosome Location:
Chr.6: 32845209 - 32853971 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00609432_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TAP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000593.5 NP_000584.2 8 - 9 2074 67
NM_001292022.1 NP_001278951.1 8 - 9 1334 67
GenBank mRNA AB012644.1 - 8 - 9 1738 67
AK123494.1 - 2 - 3 1661 67
AK130176.1 - 3 - 4 486 67
AK302340.1 - 8 - 9 1305 67
AK313762.1 - 8 - 9 2110 67
AY523970.1 - 8 - 9 1945 67
AY523971.2 - 8 - 9 1945 67
BC014081.2 - 8 - 9 2074 4304716 67
BX648013.1 - 8 - 9 2082 67
KU178346.1 - 8 - 9 1919 67
KU178347.1 - 2 - 3 464 67
L21204.1 - 8 - 9 1739 67
L21205.1 - 8 - 9 1739 67
L21206.1 - 8 - 9 1739 67
L21207.1 - 8 - 9 1739 67
L21208.1 - 8 - 9 1739 67
X57522.1 - 8 - 9 1949 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600011508.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06001574.hg.1, TC6_apd_hap1000105.hg.1, TC6_cox_hap2000199.hg.1, TC6_dbb_hap3000185.hg.1, TC6_mann_hap4000167.hg.1, TC6_mcf_hap5000174.hg.1, TC6_qbl_hap6000187.hg.1, TC6_ssto_hap7000169.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TAP1

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter

Gene Ontology Categories:

Function(s) Process(es)

adaptive immune response
antigen processing and presentation of peptide antigen via MHC class I
antigen processing and presentation of exogenous protein antigen via MHC class Ib, TAP-dependent
antigen processing and presentation of endogenous peptide antigen via MHC class I via ER pathway, TAP-dependent
antigen processing and presentation of endogenous peptide antigen via MHC class Ib via ER pathway, TAP-dependent
positive regulation of antigen processing and presentation of peptide antigen via MHC class I
defense response
peptide transport
intracellular transport of viral protein in host cell
antigen processing and presentation of endogenous peptide antigen via MHC class I
protection from natural killer cell mediated cytotoxicity
cytosol to ER transport
transmembrane transport
protein binding
ATP binding
peptide transporter activity
peptide-transporting ATPase activity
MHC class Ib protein binding
MHC class I protein binding
peptide antigen binding
ATPase activity, coupled to transmembrane movement of substances
protein homodimerization activity
ADP binding
TAP1 binding
TAP2 binding
tapasin binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000354258.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000354258.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000354258.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000354258.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000354258.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000354258.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000354258.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000354258.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000354258.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000354258.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000354258.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000354258.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000354258.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000354258.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000486332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000486332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000486332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000486332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000486332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000486332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000486332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000486332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000486332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000486332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000487296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000487296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000487296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000487296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000487296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 ENST00000487296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 NM_001292022.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 NM_001292022.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 NM_001292022.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 NM_001292022.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 NM_001292022.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 NM_001292022.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 NM_001292022.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 NM_001292022.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 NM_001292022.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 NM_001292022.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 NM_001292022.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 NM_001292022.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011508.hg.1 NM_001292022.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0