This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00610131_m1
See other STAM GE Assays ›
Gene Symbol:
STAM
Gene Name:
signal transducing adaptor molecule
Gene Aliases:
STAM-1, STAM1
Chromosome Location:
Chr.10: 17644125 - 17716824 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00610131_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: STAM
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001324282.1 NP_001311211.1 11 - 12 1331 152
NM_001324283.1 NP_001311212.1 11 - 12 1342 152
NM_001324284.1 NP_001311213.1 13 - 14 1531 152
NM_001324285.1 NP_001311214.1 13 - 14 1539 152
NM_001324286.1 NP_001311215.1 9 - 10 1170 152
NM_001324287.1 NP_001311216.1 10 - 11 1255 152
NM_001324288.1 NP_001311217.1 13 - 14 1570 152
NM_001324289.1 NP_001311218.1 12 - 13 1485 152
NM_003473.3 NP_003464.1 12 - 13 1427 152
XM_011519695.2 XP_011517997.1 12 - 13 1403 152
XM_017016685.1 XP_016872174.1 8 - 9 1472 152
GenBank mRNA AK303083.1 - 10 - 11 1115 152
AK314405.1 - 12 - 13 1311 152
U43899.1 - 12 - 13 1254 152

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000006904.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10000129.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene STAM

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
autophagy
signal transduction
positive regulation of signal transduction
endosomal transport
multivesicular body assembly
negative regulation of epidermal growth factor receptor signaling pathway
positive regulation of exosomal secretion
regulation of extracellular exosome assembly
SH3/SH2 adaptor activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR10001432.hg.1 ENST00000377524.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10001432.hg.1 ENST00000540523.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10001432.hg.1 NM_003473.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10001432.hg.1 TC10000129.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR10001433.hg.1 ENST00000377524.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10001433.hg.1 ENST00000540523.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10001433.hg.1 NM_003473.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10001433.hg.1 TC10000129.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000377500.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000377500.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000377500.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000377500.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000377500.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000377500.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000377500.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000377500.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000445846.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000445846.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000445846.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000445846.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000445846.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000445846.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000445846.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000445846.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000486183.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000486183.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000486183.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000494250.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000494250.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000494250.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 ENST00000494250.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Clariom D Assay PSR1000147257.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000147257.hg.1 NM_003473.3 Refseq
Affymetrix Human Clariom D Assay PSR1000147257.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000147257.hg.1 NM_003473.3 Refseq
Affymetrix Human Clariom D Assay PSR1000147257.hg.1 TC1000006904.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1000147258.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000147258.hg.1 NM_003473.3 Refseq
Affymetrix Human Clariom D Assay PSR1000147258.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000147258.hg.1 NM_003473.3 Refseq
Affymetrix Human Clariom D Assay PSR1000147258.hg.1 TC1000006904.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000377500.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000377500.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000377500.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000377500.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000377500.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000377500.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000377500.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000377500.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000377524.7 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000445846.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000445846.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000445846.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000445846.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000445846.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000445846.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000445846.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000445846.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000486183.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000486183.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000486183.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000494250.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000494250.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000494250.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 ENST00000494250.1 Ensembl
Affymetrix Human Clariom D Assay TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Clariom D Assay TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Clariom D Assay TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Clariom D Assay TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Clariom D Assay TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Clariom D Assay TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Clariom D Assay TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Clariom D Assay TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Clariom D Assay TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Clariom D Assay TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Clariom D Assay TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Clariom D Assay TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Clariom D Assay TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Clariom D Assay TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Clariom D Assay TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Clariom D Assay TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Clariom D Assay TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Clariom D Assay TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Clariom D Assay TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Clariom D Assay TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Clariom D Assay TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Clariom D Assay TC1000006904.hg.1 NM_003473.3 Refseq
Affymetrix Human Clariom D Assay TC1000006904.hg.1 NM_003473.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline