This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00611072_g1
See other LMF2 GE Assays ›
Gene Symbol:
LMF2
Gene Name:
lipase maturation factor 2
Gene Aliases:
TMEM112B, TMEM153
Chromosome Location:
Chr.22: 50502947 - 50507840 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00611072_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LMF2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_033200.2 NP_149977.2 7 - 8 1077 75
XM_005261955.3 XP_005262012.1 7 - 8 1159 75
XM_006724426.3 XP_006724489.1 6 - 7 1523 75
XM_006724427.3 XP_006724490.1 7 - 8 1074 75
GenBank mRNA AK130818.1 - 1 - 2 227 75
BC002942.2 - 7 - 8 1051 3954961 75
BC014652.1 - 7 - 8 1140 4562676 75
BC021143.1 - 7 - 8 1051 4872642 75
CR456605.1 - 7 - 8 1061 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2200009161.hg.1
This assay targets the same location as HTA transcript cluster(s) TC22000914.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene LMF2

Gene Ontology Categories:

Process(es)

protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR22015593.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22015593.hg.1 ENST00000380796.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22015593.hg.1 ENST00000474879.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22015593.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22015593.hg.1 TC22000914.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000505981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000505981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000505981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000505981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000505981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000505981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000507607.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000507607.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000507607.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000507607.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000507607.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay PSR2200166499.hg.1 TC2200009161.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2200166499.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200166499.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay PSR2200166499.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Clariom D Assay PSR2200166499.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay PSR2200166499.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200166499.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay PSR2200166499.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Clariom D Assay PSR2200166499.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000505981.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000505981.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000505981.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000505981.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000505981.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000505981.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000507607.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000507607.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000507607.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000507607.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000507607.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0