This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00698272_m1
See other PDE12 GE Assays ›
Gene Symbol:
PDE12
Gene Name:
phosphodiesterase 12
Gene Aliases:
2'-PDE, 2-PDE
Chromosome Location:
Chr.3: 57556247 - 57656480 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00698272_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer:
284.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PDE12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001322176.1 NP_001309105.1 1 - 2 1441 114
NM_001322177.1 NP_001309106.1 1 - 2 1441 114
NM_177966.6 NP_808881.3 1 - 2 1441 114
GenBank mRNA AB115695.1 - 1 - 2 1379 114
AK074423.1 - 1 - 2 1390 114
AK300374.1 - 1 - 2 1414 114
AL831824.1 - 1 - 2 945 114
BM713922.1 - 1 - 2 464 114
BX647169.1 - 1 - 2 1411 114

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PDE12

Panther Classification:

Molecular Function -

mRNA polyadenylation factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
RNA phosphodiester bond hydrolysis, exonucleolytic
poly(A)-specific ribonuclease activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300007579.hg.1 ENST00000311180.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007579.hg.1 ENST00000311180.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007579.hg.1 ENST00000311180.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007579.hg.1 ENST00000311180.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007579.hg.1 ENST00000311180.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007579.hg.1 ENST00000311180.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007579.hg.1 ENST00000311180.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007579.hg.1 ENST00000487257.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007579.hg.1 ENST00000487257.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007579.hg.1 ENST00000487257.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007579.hg.1 ENST00000487257.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007579.hg.1 ENST00000487257.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007579.hg.1 ENST00000487257.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007579.hg.1 NM_177966.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007579.hg.1 NM_177966.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007579.hg.1 NM_177966.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007579.hg.1 NM_177966.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007579.hg.1 NM_177966.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007579.hg.1 NM_177966.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007584.hg.1 ENST00000470427.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007584.hg.1 ENST00000470427.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007584.hg.1 ENST00000470427.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007584.hg.1 ENST00000470427.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007579.hg.1 ENST00000311180.8 Ensembl
Affymetrix Human Clariom D Assay TC0300007579.hg.1 ENST00000311180.8 Ensembl
Affymetrix Human Clariom D Assay TC0300007579.hg.1 ENST00000311180.8 Ensembl
Affymetrix Human Clariom D Assay TC0300007579.hg.1 ENST00000311180.8 Ensembl
Affymetrix Human Clariom D Assay TC0300007579.hg.1 ENST00000311180.8 Ensembl
Affymetrix Human Clariom D Assay TC0300007579.hg.1 ENST00000311180.8 Ensembl
Affymetrix Human Clariom D Assay TC0300007579.hg.1 ENST00000311180.8 Ensembl
Affymetrix Human Clariom D Assay TC0300007579.hg.1 ENST00000487257.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007579.hg.1 ENST00000487257.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007579.hg.1 ENST00000487257.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007579.hg.1 ENST00000487257.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007579.hg.1 ENST00000487257.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007579.hg.1 ENST00000487257.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007579.hg.1 NM_177966.6 Refseq
Affymetrix Human Clariom D Assay TC0300007579.hg.1 NM_177966.6 Refseq
Affymetrix Human Clariom D Assay TC0300007579.hg.1 NM_177966.6 Refseq
Affymetrix Human Clariom D Assay TC0300007579.hg.1 NM_177966.6 Refseq
Affymetrix Human Clariom D Assay TC0300007579.hg.1 NM_177966.6 Refseq
Affymetrix Human Clariom D Assay TC0300007579.hg.1 NM_177966.6 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline