This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00698323_g1
See other MOGAT3 GE Assays ›
Gene Symbol:
MOGAT3
Gene Name:
monoacylglycerol O-acyltransferase 3
Gene Aliases:
DC7, DGAT2L2, MGAT3
Chromosome Location:
Chr.7: 101192886 - 101201131 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00698323_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MOGAT3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001287147.1 NP_001274076.1 1 - 2 279 73
NM_178176.3 NP_835470.1 1 - 2 279 73
XM_005250309.3 XP_005250366.1 1 - 2 389 73
GenBank mRNA AY229854.1 - 1 - 2 279 73
AY358200.1 - 1 - 2 256 73
BC100953.1 - 1 - 2 170 40004561 73
BC100954.1 - 1 - 2 170 40004562 73
BC100955.1 - 1 - 2 173 40004564 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700012038.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07001685.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MOGAT3

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

glycerol metabolic process
triglyceride biosynthetic process
2-acylglycerol O-acyltransferase activity
diacylglycerol O-acyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 ENST00000223114.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 ENST00000223114.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 ENST00000223114.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 ENST00000223114.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 ENST00000223114.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 ENST00000223114.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 ENST00000223114.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 ENST00000223114.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 ENST00000223114.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 ENST00000223114.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 ENST00000223114.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 ENST00000223114.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 ENST00000379423.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 ENST00000379423.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 ENST00000379423.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 ENST00000379423.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 ENST00000379423.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 ENST00000379423.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 ENST00000379423.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 ENST00000379423.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 ENST00000440203.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 ENST00000440203.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 ENST00000440203.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 ENST00000440203.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 ENST00000440203.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 ENST00000440203.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 ENST00000440203.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 ENST00000440203.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 ENST00000440203.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 ENST00000440203.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 NM_001287147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 NM_001287147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 NM_001287147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 NM_001287147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 NM_001287147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 NM_001287147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 NM_001287147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 NM_001287147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 NM_001287147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 NM_001287147.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 NM_178176.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 NM_178176.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 NM_178176.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 NM_178176.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 NM_178176.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 NM_178176.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 NM_178176.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 NM_178176.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 NM_178176.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 NM_178176.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012038.hg.1 NM_178176.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07026048.hg.1 TC07001685.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR07026048.hg.1 ENST00000223114.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07026048.hg.1 ENST00000379423.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07026048.hg.1 ENST00000440203.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07026048.hg.1 NM_178176.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07026047.hg.1 TC07001685.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR07026047.hg.1 ENST00000223114.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07026047.hg.1 ENST00000379423.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07026047.hg.1 ENST00000440203.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07026047.hg.1 NM_178176.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07026050.hg.1 TC07001685.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR07026050.hg.1 ENST00000223114.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07026050.hg.1 NM_178176.3 Refseq
Affymetrix Human Clariom D Assay TC0700012038.hg.1 ENST00000223114.8 Ensembl
Affymetrix Human Clariom D Assay TC0700012038.hg.1 ENST00000223114.8 Ensembl
Affymetrix Human Clariom D Assay TC0700012038.hg.1 ENST00000223114.8 Ensembl
Affymetrix Human Clariom D Assay TC0700012038.hg.1 ENST00000223114.8 Ensembl
Affymetrix Human Clariom D Assay TC0700012038.hg.1 ENST00000223114.8 Ensembl
Affymetrix Human Clariom D Assay TC0700012038.hg.1 ENST00000223114.8 Ensembl
Affymetrix Human Clariom D Assay TC0700012038.hg.1 ENST00000223114.8 Ensembl
Affymetrix Human Clariom D Assay TC0700012038.hg.1 ENST00000223114.8 Ensembl
Affymetrix Human Clariom D Assay TC0700012038.hg.1 ENST00000223114.8 Ensembl
Affymetrix Human Clariom D Assay TC0700012038.hg.1 ENST00000223114.8 Ensembl
Affymetrix Human Clariom D Assay TC0700012038.hg.1 ENST00000223114.8 Ensembl
Affymetrix Human Clariom D Assay TC0700012038.hg.1 ENST00000223114.8 Ensembl
Affymetrix Human Clariom D Assay TC0700012038.hg.1 ENST00000379423.3 Ensembl
Affymetrix Human Clariom D Assay TC0700012038.hg.1 ENST00000379423.3 Ensembl
Affymetrix Human Clariom D Assay TC0700012038.hg.1 ENST00000379423.3 Ensembl
Affymetrix Human Clariom D Assay TC0700012038.hg.1 ENST00000379423.3 Ensembl
Affymetrix Human Clariom D Assay TC0700012038.hg.1 ENST00000379423.3 Ensembl
Affymetrix Human Clariom D Assay TC0700012038.hg.1 ENST00000379423.3 Ensembl
Affymetrix Human Clariom D Assay TC0700012038.hg.1 ENST00000379423.3 Ensembl
Affymetrix Human Clariom D Assay TC0700012038.hg.1 ENST00000379423.3 Ensembl
Affymetrix Human Clariom D Assay TC0700012038.hg.1 ENST00000440203.6 Ensembl
Affymetrix Human Clariom D Assay TC0700012038.hg.1 ENST00000440203.6 Ensembl
Affymetrix Human Clariom D Assay TC0700012038.hg.1 ENST00000440203.6 Ensembl
Affymetrix Human Clariom D Assay TC0700012038.hg.1 ENST00000440203.6 Ensembl
Affymetrix Human Clariom D Assay TC0700012038.hg.1 ENST00000440203.6 Ensembl
Affymetrix Human Clariom D Assay TC0700012038.hg.1 ENST00000440203.6 Ensembl
Affymetrix Human Clariom D Assay TC0700012038.hg.1 ENST00000440203.6 Ensembl
Affymetrix Human Clariom D Assay TC0700012038.hg.1 ENST00000440203.6 Ensembl
Affymetrix Human Clariom D Assay TC0700012038.hg.1 ENST00000440203.6 Ensembl
Affymetrix Human Clariom D Assay TC0700012038.hg.1 ENST00000440203.6 Ensembl
Affymetrix Human Clariom D Assay TC0700012038.hg.1 NM_001287147.1 Refseq
Affymetrix Human Clariom D Assay TC0700012038.hg.1 NM_001287147.1 Refseq
Affymetrix Human Clariom D Assay TC0700012038.hg.1 NM_001287147.1 Refseq
Affymetrix Human Clariom D Assay TC0700012038.hg.1 NM_001287147.1 Refseq
Affymetrix Human Clariom D Assay TC0700012038.hg.1 NM_001287147.1 Refseq
Affymetrix Human Clariom D Assay TC0700012038.hg.1 NM_001287147.1 Refseq
Affymetrix Human Clariom D Assay TC0700012038.hg.1 NM_001287147.1 Refseq
Affymetrix Human Clariom D Assay TC0700012038.hg.1 NM_001287147.1 Refseq
Affymetrix Human Clariom D Assay TC0700012038.hg.1 NM_001287147.1 Refseq
Affymetrix Human Clariom D Assay TC0700012038.hg.1 NM_001287147.1 Refseq
Affymetrix Human Clariom D Assay TC0700012038.hg.1 NM_178176.3 Refseq
Affymetrix Human Clariom D Assay TC0700012038.hg.1 NM_178176.3 Refseq
Affymetrix Human Clariom D Assay TC0700012038.hg.1 NM_178176.3 Refseq
Affymetrix Human Clariom D Assay TC0700012038.hg.1 NM_178176.3 Refseq
Affymetrix Human Clariom D Assay TC0700012038.hg.1 NM_178176.3 Refseq
Affymetrix Human Clariom D Assay TC0700012038.hg.1 NM_178176.3 Refseq
Affymetrix Human Clariom D Assay TC0700012038.hg.1 NM_178176.3 Refseq
Affymetrix Human Clariom D Assay TC0700012038.hg.1 NM_178176.3 Refseq
Affymetrix Human Clariom D Assay TC0700012038.hg.1 NM_178176.3 Refseq
Affymetrix Human Clariom D Assay TC0700012038.hg.1 NM_178176.3 Refseq
Affymetrix Human Clariom D Assay TC0700012038.hg.1 NM_178176.3 Refseq
Affymetrix Human Clariom D Assay PSR0700186519.hg.1 ENST00000223114.8 Ensembl
Affymetrix Human Clariom D Assay PSR0700186519.hg.1 ENST00000379423.3 Ensembl
Affymetrix Human Clariom D Assay PSR0700186519.hg.1 ENST00000440203.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700186519.hg.1 NM_001287147.1 Refseq
Affymetrix Human Clariom D Assay PSR0700186519.hg.1 NM_178176.3 Refseq
Affymetrix Human Clariom D Assay PSR0700186519.hg.1 ENST00000223114.8 Ensembl
Affymetrix Human Clariom D Assay PSR0700186519.hg.1 ENST00000379423.3 Ensembl
Affymetrix Human Clariom D Assay PSR0700186519.hg.1 ENST00000440203.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700186519.hg.1 NM_001287147.1 Refseq
Affymetrix Human Clariom D Assay PSR0700186519.hg.1 NM_178176.3 Refseq
Affymetrix Human Clariom D Assay PSR0700186519.hg.1 TC0700012038.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0700186520.hg.1 ENST00000223114.8 Ensembl
Affymetrix Human Clariom D Assay PSR0700186520.hg.1 ENST00000379423.3 Ensembl
Affymetrix Human Clariom D Assay PSR0700186520.hg.1 ENST00000440203.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700186520.hg.1 NM_001287147.1 Refseq
Affymetrix Human Clariom D Assay PSR0700186520.hg.1 NM_178176.3 Refseq
Affymetrix Human Clariom D Assay PSR0700186520.hg.1 ENST00000223114.8 Ensembl
Affymetrix Human Clariom D Assay PSR0700186520.hg.1 ENST00000379423.3 Ensembl
Affymetrix Human Clariom D Assay PSR0700186520.hg.1 ENST00000440203.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700186520.hg.1 NM_001287147.1 Refseq
Affymetrix Human Clariom D Assay PSR0700186520.hg.1 NM_178176.3 Refseq
Affymetrix Human Clariom D Assay PSR0700186520.hg.1 TC0700012038.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0700186522.hg.1 ENST00000223114.8 Ensembl
Affymetrix Human Clariom D Assay PSR0700186522.hg.1 NM_001287147.1 Refseq
Affymetrix Human Clariom D Assay PSR0700186522.hg.1 NM_178176.3 Refseq
Affymetrix Human Clariom D Assay PSR0700186522.hg.1 ENST00000223114.8 Ensembl
Affymetrix Human Clariom D Assay PSR0700186522.hg.1 NM_001287147.1 Refseq
Affymetrix Human Clariom D Assay PSR0700186522.hg.1 NM_178176.3 Refseq
Affymetrix Human Clariom D Assay PSR0700186522.hg.1 TC0700012038.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0700186518.hg.1 ENST00000223114.8 Ensembl
Affymetrix Human Clariom D Assay PSR0700186518.hg.1 ENST00000379423.3 Ensembl
Affymetrix Human Clariom D Assay PSR0700186518.hg.1 ENST00000440203.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700186518.hg.1 NM_001287147.1 Refseq
Affymetrix Human Clariom D Assay PSR0700186518.hg.1 NM_178176.3 Refseq
Affymetrix Human Clariom D Assay PSR0700186518.hg.1 ENST00000223114.8 Ensembl
Affymetrix Human Clariom D Assay PSR0700186518.hg.1 ENST00000379423.3 Ensembl
Affymetrix Human Clariom D Assay PSR0700186518.hg.1 ENST00000440203.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700186518.hg.1 NM_001287147.1 Refseq
Affymetrix Human Clariom D Assay PSR0700186518.hg.1 NM_178176.3 Refseq
Affymetrix Human Clariom D Assay PSR0700186518.hg.1 TC0700012038.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0