This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00699131_g1
See other GRASP GE Assays ›
Gene Symbol:
GRASP
Gene Name:
general receptor for phosphoinositides 1 associated scaffold protein
Gene Aliases:
TAMALIN
Chromosome Location:
Chr.12: 52006945 - 52015889 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00699131_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GRASP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001271856.1 NP_001258785.1 4 - 5 391 67
NM_181711.3 NP_859062.1 5 - 6 625 67
XM_005268691.3 XP_005268748.1 5 - 6 1793 67
XM_011537996.1 XP_011536298.1 4 - 5 1571 67
GenBank mRNA BC035500.2 - 4 - 5 362 4422992 67
BC052967.1 - 4 - 5 309 67
BC053666.1 - 5 - 6 606 6151168 67
HY035281.1 - 4 - 5 325 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200007652.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12000413.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GRASP

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
protein localization
PDZ domain binding
ADP-ribosylation factor binding
identical protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR12005282.hg.1 ENST00000293662.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12005282.hg.1 ENST00000380039.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12005282.hg.1 ENST00000552049.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12005282.hg.1 ENST00000552963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12005282.hg.1 NM_181711.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12005282.hg.1 TC12000413.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000293662.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000293662.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000293662.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000293662.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000293662.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000293662.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000293662.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000293662.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000293662.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000293662.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000293662.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000293662.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000293662.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000546756.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000546756.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000546756.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000546756.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000546756.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000546756.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000546756.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000546756.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000546756.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000547749.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000547749.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000547749.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000548505.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000548505.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000548505.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000548505.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000548505.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000550809.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000550809.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000550809.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000550809.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000550809.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000550809.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000552049.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000552049.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000552049.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000552049.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000552049.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000552049.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000552049.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000552049.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000552049.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000552049.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000552049.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000552049.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000552049.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000552508.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000552508.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000552508.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000552508.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000552508.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000552508.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000552963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000552963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000552963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000552963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000552963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000552963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000552963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000552963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000552963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000552963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 ENST00000552963.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 NM_001271856.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 NM_001271856.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 NM_001271856.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 NM_001271856.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 NM_001271856.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 NM_001271856.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 NM_001271856.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 NM_001271856.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 NM_001271856.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 NM_001271856.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 NM_001271856.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 NM_001271856.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 NM_001271856.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 NM_001271856.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 NM_181711.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 NM_181711.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 NM_181711.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 NM_181711.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 NM_181711.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 NM_181711.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 NM_181711.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 NM_181711.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 NM_181711.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 NM_181711.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 NM_181711.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 NM_181711.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007652.hg.1 NM_181711.3 Refseq
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000293662.8 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000293662.8 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000293662.8 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000293662.8 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000293662.8 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000293662.8 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000293662.8 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000293662.8 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000293662.8 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000293662.8 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000293662.8 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000293662.8 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000293662.8 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000546756.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000546756.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000546756.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000546756.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000546756.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000546756.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000546756.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000546756.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000546756.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000547749.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000547749.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000547749.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000548505.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000548505.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000548505.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000548505.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000548505.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000550809.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000550809.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000550809.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000550809.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000550809.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000550809.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000552049.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000552049.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000552049.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000552049.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000552049.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000552049.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000552049.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000552049.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000552049.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000552049.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000552049.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000552049.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000552049.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000552508.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000552508.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000552508.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000552508.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000552508.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000552508.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000552963.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000552963.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000552963.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000552963.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000552963.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000552963.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000552963.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000552963.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000552963.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000552963.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 ENST00000552963.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007652.hg.1 NM_001271856.1 Refseq
Affymetrix Human Clariom D Assay TC1200007652.hg.1 NM_001271856.1 Refseq
Affymetrix Human Clariom D Assay TC1200007652.hg.1 NM_001271856.1 Refseq
Affymetrix Human Clariom D Assay TC1200007652.hg.1 NM_001271856.1 Refseq
Affymetrix Human Clariom D Assay TC1200007652.hg.1 NM_001271856.1 Refseq
Affymetrix Human Clariom D Assay TC1200007652.hg.1 NM_001271856.1 Refseq
Affymetrix Human Clariom D Assay TC1200007652.hg.1 NM_001271856.1 Refseq
Affymetrix Human Clariom D Assay TC1200007652.hg.1 NM_001271856.1 Refseq
Affymetrix Human Clariom D Assay TC1200007652.hg.1 NM_001271856.1 Refseq
Affymetrix Human Clariom D Assay TC1200007652.hg.1 NM_001271856.1 Refseq
Affymetrix Human Clariom D Assay TC1200007652.hg.1 NM_001271856.1 Refseq
Affymetrix Human Clariom D Assay TC1200007652.hg.1 NM_001271856.1 Refseq
Affymetrix Human Clariom D Assay TC1200007652.hg.1 NM_001271856.1 Refseq
Affymetrix Human Clariom D Assay TC1200007652.hg.1 NM_001271856.1 Refseq
Affymetrix Human Clariom D Assay TC1200007652.hg.1 NM_181711.3 Refseq
Affymetrix Human Clariom D Assay TC1200007652.hg.1 NM_181711.3 Refseq
Affymetrix Human Clariom D Assay TC1200007652.hg.1 NM_181711.3 Refseq
Affymetrix Human Clariom D Assay TC1200007652.hg.1 NM_181711.3 Refseq
Affymetrix Human Clariom D Assay TC1200007652.hg.1 NM_181711.3 Refseq
Affymetrix Human Clariom D Assay TC1200007652.hg.1 NM_181711.3 Refseq
Affymetrix Human Clariom D Assay TC1200007652.hg.1 NM_181711.3 Refseq
Affymetrix Human Clariom D Assay TC1200007652.hg.1 NM_181711.3 Refseq
Affymetrix Human Clariom D Assay TC1200007652.hg.1 NM_181711.3 Refseq
Affymetrix Human Clariom D Assay TC1200007652.hg.1 NM_181711.3 Refseq
Affymetrix Human Clariom D Assay TC1200007652.hg.1 NM_181711.3 Refseq
Affymetrix Human Clariom D Assay TC1200007652.hg.1 NM_181711.3 Refseq
Affymetrix Human Clariom D Assay TC1200007652.hg.1 NM_181711.3 Refseq
Affymetrix Human Clariom D Assay PSR1200154487.hg.1 ENST00000550809.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200154487.hg.1 ENST00000552049.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200154487.hg.1 NM_001271856.1 Refseq
Affymetrix Human Clariom D Assay PSR1200154487.hg.1 ENST00000550809.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200154487.hg.1 ENST00000552049.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200154487.hg.1 NM_001271856.1 Refseq
Affymetrix Human Clariom D Assay PSR1200154487.hg.1 TC1200007652.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1200154494.hg.1 ENST00000293662.8 Ensembl
Affymetrix Human Clariom D Assay PSR1200154494.hg.1 ENST00000546756.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200154494.hg.1 ENST00000548505.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200154494.hg.1 ENST00000552049.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200154494.hg.1 ENST00000552508.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200154494.hg.1 ENST00000552963.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200154494.hg.1 NM_001271856.1 Refseq
Affymetrix Human Clariom D Assay PSR1200154494.hg.1 NM_181711.3 Refseq
Affymetrix Human Clariom D Assay PSR1200154494.hg.1 ENST00000293662.8 Ensembl
Affymetrix Human Clariom D Assay PSR1200154494.hg.1 ENST00000546756.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200154494.hg.1 ENST00000548505.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200154494.hg.1 ENST00000552049.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200154494.hg.1 ENST00000552508.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200154494.hg.1 ENST00000552963.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200154494.hg.1 NM_001271856.1 Refseq
Affymetrix Human Clariom D Assay PSR1200154494.hg.1 NM_181711.3 Refseq
Affymetrix Human Clariom D Assay PSR1200154494.hg.1 TC1200007652.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0